Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other AP1G2 SIRNA Assays ›
          Gene Symbol:
          AP1G2
          Gene Name:
          adaptor related protein complex 1 gamma 2 subunit
          Gene Aliases:
          G2AD
          Chromosome Location:
          Chr.14: 23559563 - 23569056 on Build GRCh38
          UniGene ID:
          Hs.343244
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS144924
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: AP1G2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001282474.1 NP_001269403.1 7, 8 1074
          NM_001282475.1 NP_001269404.1 7, 8 924
          NM_003917.4 NP_003908.1 9, 10 1028
          XM_005268167.3 XP_005268224.1 8, 9 1151
          XM_005268168.4 XP_005268225.1 8, 9 1193
          XM_005268169.3 XP_005268226.1 8, 9 1150
          XM_005268170.3 XP_005268227.1 8, 9 1150
          XM_005268172.3 XP_005268229.1 9, 10 979
          XM_005268173.3 XP_005268230.1 9, 10 979
          XM_005268174.3 XP_005268231.1 7, 8 1026
          XM_005268175.3 XP_005268232.1 7, 8 1040
          XM_005268177.3 XP_005268234.1 8, 9 1006
          XM_005268178.3 XP_005268235.1 8, 9 897
          XM_005268179.3 XP_005268236.1 8, 9 1002
          XM_005268180.4 XP_005268237.1 7, 8 1244
          XM_005268182.3 XP_005268239.1 8, 9 873
          XM_006720301.3 XP_006720364.1 8, 9 1248
          XM_011537283.2 XP_011535585.1 7, 8 1097
          XM_011537286.2 XP_011535588.1 1, 2 21
          XM_017021740.1 XP_016877229.1 7, 8 1097
          XM_017021741.1 XP_016877230.1 5, 6 1001
          XM_017021742.1 XP_016877231.1 7, 8 1068
          XM_017021743.1 XP_016877232.1 7, 8 1097
          XM_017021744.1 XP_016877233.1 9, 10 980
          XM_017021745.1 XP_016877234.1 9, 10 980
          XM_017021746.1 XP_016877235.1 8, 9 873
          XM_017021747.1 XP_016877236.1 8, 9 2265
          XM_017021748.1 XP_016877237.1 7, 8 766
          GenBank mRNA AB015318.1 - 9, 10 970
          AF068706.1 - 8, 9 1688
          AK123354.1 - 1, 2 75
          AK304682.1 - 7, 8 759
          BC051833.1 - 6, 7 865 5589077
          BX161425.1 - 7, 8 1007
          BX247963.1 - 6, 7 733

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          membrane traffic protein

          Gene Ontology Categories:

          Function(s) Process(es)

          intracellular protein transport
          viral process
          vesicle-mediated transport
          protein binding
          protein transporter activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-66d58:80/100.66.79.157:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0