Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RBM39 SIRNA Assays ›
Gene Symbol:
RBM39
Gene Name:
RNA binding motif protein 39
Gene Aliases:
CAPER, CAPERalpha, FSAP59, HCC1, RNPC2
Chromosome Location:
Chr.20: 35703609 - 35742336 on Build GRCh38
UniGene ID:
Hs.282901
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS145210
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RBM39
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242599.1
NP_001229528.1
8, 9
1174
NM_001242600.1
NP_001229529.1
8, 9
1174
NM_001323422.1
NP_001310351.1
9, 10
1240
NM_001323423.1
NP_001310352.1
11, 12
1385
NM_001323424.1
NP_001310353.1
9, 10
1237
NM_004902.3
NP_004893.1
9, 10
1240
NM_184234.2
NP_909122.1
9, 10
1240
NR_040722.1
10, 11
1313
NR_040723.1
10, 11
1312
NR_040724.1
10, 11
1313
XM_006723890.3
XP_006723953.1
9, 10
2820
XM_006723891.3
XP_006723954.1
10, 11
1843
XM_011529111.2
XP_011527413.1
10, 11
2892
XM_017028138.1
XP_016883627.1
8, 9
1174
XM_017028139.1
XP_016883628.1
11, 12
2209
XM_017028140.1
XP_016883629.1
9, 10
1247
XM_017028141.1
XP_016883630.1
9, 10
2820
XM_017028142.1
XP_016883631.1
10, 11
3716
XM_017028143.1
XP_016883632.1
10, 11
2892
XM_017028144.1
XP_016883633.1
11, 12
1385
XM_017028145.1
XP_016883634.1
9, 10
1247
XM_017028146.1
XP_016883635.1
10, 11
1313
XM_017028147.1
XP_016883636.1
10, 11
1236
GenBank mRNA
AK126158.1
-
6, 7
1399
AK297520.1
-
4, 5
611
AK299165.1
-
8, 9
1074
AK299678.1
-
8, 9
1076
AK303910.1
-
9, 10
1112
BC131543.1
-
8, 9
940
40085367
BC141835.1
-
9, 10
1090
40148871
BC158172.1
-
9, 10
1090
40148190
BX537770.1
-
10, 11
1232
BX640714.1
-
10, 11
1225
BX640812.1
-
10, 11
1222
CR749443.1
-
9, 10
2907
L10910.1
-
9, 10
980
L10911.1
-
8, 9
855
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA splicing factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
RNA processing
mRNA processing
RNA splicing
nucleotide binding
protein binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0