Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BCL2L11 SIRNA Assays ›
Gene Symbol:
BCL2L11
Gene Name:
BCL2 like 11
Gene Aliases:
BAM, BIM, BOD
Chromosome Location:
Chr.2: 111120914 - 111168445 on Build GRCh38
UniGene ID:
Hs.469658
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS145413
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BCL2L11
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001204106.1
NP_001191035.1
2
328
NM_001204107.1
NP_001191036.1
2
328
NM_001204108.1
NP_001191037.1
2
328
NM_001204109.1
NP_001191038.1
2
328
NM_001204110.1
NP_001191039.1
2
328
NM_001204111.1
NP_001191040.1
2
328
NM_001204112.1
NP_001191041.1
2
328
NM_001204113.1
NP_001191042.1
2
328
NM_006538.4
NP_006529.1
2
328
NM_138621.4
NP_619527.1
2
328
NM_138622.3
NP_619528.1
2
328
NM_138623.3
NP_619529.1
2
328
NM_138624.3
NP_619530.1
2
328
NM_138625.3
NP_619531.1
2
328
NM_138626.3
NP_619532.1
2
328
NM_138627.3
NP_619533.1
2
328
NM_207002.3
NP_996885.1
2
328
NM_207003.2
NP_996886.1
2
328
XM_005263550.3
XP_005263607.1
2
1106
XM_005263551.2
XP_005263608.1
2
1106
XM_005263552.3
XP_005263609.1
2
1106
XM_005263553.2
XP_005263610.1
2
1106
XM_005263554.2
XP_005263611.1
2
1106
XM_005263555.3
XP_005263612.1
2
1106
XM_005263556.3
XP_005263613.1
2
328
XM_005263557.4
XP_005263614.1
2
405
XM_005263559.2
XP_005263616.1
2
1106
XM_005263561.3
XP_005263618.1
2
328
XM_011510461.2
XP_011508763.1
2
1106
XM_011510464.2
XP_011508766.1
2
1106
XM_017003101.1
XP_016858590.1
2
1106
XM_017003102.1
XP_016858591.1
2
328
GenBank mRNA
AB071195.1
-
1
40
AB071196.1
-
1
40
AB071197.1
-
1
40
AB071198.1
-
1
40
AB071199.1
-
1
40
AB071200.1
-
1
40
AF032457.1
-
1
40
AF032458.1
-
1
40
AK290377.1
-
2
328
AK291269.1
-
2
325
AY305714.1
-
1
40
AY305715.1
-
1
40
AY352518.1
-
1
40
AY423441.1
-
1
40
AY423442.1
-
1
40
AY423443.1
-
1
40
AY428962.1
-
1
40
BC033694.1
-
2
310
5213713
CN411912.1
-
2
297
DQ849200.1
-
1
40
DQ849201.1
-
1
40
DQ849202.1
-
1
40
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
in utero embryonic development
B cell homeostasis
B cell apoptotic process
kidney development
protein insertion into mitochondrial membrane involved in apoptotic signaling pathway
myeloid cell homeostasis
apoptotic process
meiosis I
cell-matrix adhesion
spermatogenesis
brain development
male gonad development
intrinsic apoptotic signaling pathway in response to DNA damage
mammary gland development
positive regulation of protein homooligomerization
response to endoplasmic reticulum stress
tube formation
odontogenesis of dentin-containing tooth
T cell homeostasis
positive regulation of apoptotic process
positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
positive regulation of neuron apoptotic process
ear development
positive regulation of cell cycle
regulation of organ growth
developmental pigmentation
regulation of developmental pigmentation
spleen development
thymus development
post-embryonic organ morphogenesis
positive regulation of apoptotic process by virus
cellular process regulating host cell cycle in response to virus
thymocyte apoptotic process
positive regulation of release of cytochrome c from mitochondria
extrinsic apoptotic signaling pathway in absence of ligand
positive regulation of protein insertion into mitochondrial membrane involved in apoptotic signaling pathway
positive regulation of endoplasmic reticulum stress-induced intrinsic apoptotic signaling pathway
apoptotic process involved in embryonic digit morphogenesis
positive regulation of IRE1-mediated unfolded protein response
positive regulation of fibroblast apoptotic process
positive regulation of intrinsic apoptotic signaling pathway
protein binding
microtubule binding
dynein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0