Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ARPP21 SIRNA Assays ›
Gene Symbol:
ARPP21
Gene Name:
cAMP regulated phosphoprotein 21
Gene Aliases:
ARPP-21, R3HDM3, RCS, TARPP
Chromosome Location:
Chr.3: 35638981 - 35798933 on Build GRCh38
UniGene ID:
Hs.475902
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS145793
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ARPP21
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001025068.1
NP_001020239.1
3, 4
420
NM_001025069.1
NP_001020240.1
3, 4
429
NM_001267616.1
NP_001254545.1
3, 4
1576
NM_001267617.1
NP_001254546.1
3, 4
386
NM_001267618.1
NP_001254547.1
2, 3
517
NM_001267619.1
NP_001254548.1
2, 3
351
NM_016300.4
NP_057384.2
3, 4
403
NM_198399.1
NP_938409.1
2, 3
411
XM_005264811.2
XP_005264868.1
3, 4
432
XM_006712943.2
XP_006713006.1
3, 4
432
XM_006712944.2
XP_006713007.1
3, 4
432
XM_011533298.2
XP_011531600.1
3, 4
432
XM_011533299.2
XP_011531601.1
3, 4
976
XM_011533300.2
XP_011531602.1
3, 4
595
XM_011533301.2
XP_011531603.1
3, 4
426
XM_011533302.2
XP_011531604.1
4, 5
518
XM_011533303.2
XP_011531605.1
4, 5
1068
XM_017005574.1
XP_016861063.1
3, 4
522
XM_017005575.1
XP_016861064.1
4, 5
12155
XM_017005576.1
XP_016861065.1
3, 4
976
XM_017005577.1
XP_016861066.1
3, 4
426
XM_017005578.1
XP_016861067.1
3, 4
402
XM_017005579.1
XP_016861068.1
3, 4
976
XM_017005580.1
XP_016861069.1
4, 5
1068
XM_017005581.1
XP_016861070.1
4, 5
12155
XM_017005582.1
XP_016861071.1
3, 4
976
XM_017005583.1
XP_016861072.1
3, 4
426
XM_017005584.1
XP_016861073.1
3, 4
976
XM_017005585.1
XP_016861074.1
3, 4
976
XM_017005586.1
XP_016861075.1
3, 4
432
XM_017005587.1
XP_016861076.1
3, 4
595
XM_017005588.1
XP_016861077.1
4, 5
1068
XM_017005589.1
XP_016861078.1
4, 5
518
XM_017005590.1
XP_016861079.1
4, 5
12155
XM_017005591.1
XP_016861080.1
3, 4
976
XM_017005592.1
XP_016861081.1
3, 4
976
XM_017005593.1
XP_016861082.1
3, 4
522
XM_017005594.1
XP_016861083.1
3, 4
426
XM_017005595.1
XP_016861084.1
4, 5
1068
XM_017005596.1
XP_016861085.1
3, 4
976
XM_017005597.1
XP_016861086.1
3, 4
976
XM_017005598.1
XP_016861087.1
4, 5
1068
XM_017005599.1
XP_016861088.1
3, 4
976
XM_017005600.1
XP_016861089.1
3, 4
595
XM_017005601.1
XP_016861090.1
3, 4
426
XM_017005602.1
XP_016861091.1
3, 4
976
XM_017005603.1
XP_016861092.1
3, 4
595
XM_017005604.1
XP_016861093.1
3, 4
426
XM_017005605.1
XP_016861094.1
4, 5
1068
XM_017005606.1
XP_016861095.1
4, 5
518
XM_017005607.1
XP_016861096.1
3, 4
976
XM_017005608.1
XP_016861097.1
3, 4
976
XM_017005609.1
XP_016861098.1
4, 5
1068
XM_017005610.1
XP_016861099.1
3, 4
976
XM_017005611.1
XP_016861100.1
3, 4
976
XM_017005612.1
XP_016861101.1
3, 4
976
GenBank mRNA
AF112220.1
-
2, 3
376
AK055064.1
-
3, 4
429
AK289649.1
-
2, 3
366
AK289868.1
-
3, 4
423
AK294484.1
-
2, 3
364
AV718064.1
-
3, 4
420
AV718166.1
-
3, 4
429
BC017805.1
-
2, 3
411
BC036399.1
-
3, 4
391
5263894
BC041385.1
-
3, 4
1164
BC051828.1
-
3, 4
403
6012704
BC066651.1
-
1, 2
930
BG535216.1
-
2, 3
412
DA275579.1
-
2, 3
517
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
biological_process
cellular response to heat
molecular_function
nucleic acid binding
calmodulin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0