Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MACF1 SIRNA Assays ›
Gene Symbol:
MACF1
Gene Name:
microtubule-actin crosslinking factor 1
Gene Aliases:
ABP620, ACF7, MACF, OFC4
Chromosome Location:
Chr.1: 39084167 - 39487138 on Build GRCh38
UniGene ID:
Hs.472475
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS146438
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MACF1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_012090.5
NP_036222.3
74
13241
XM_005270694.1
XP_005270751.1
75
20103
XM_005270695.1
XP_005270752.1
75
20103
XM_005270696.1
XP_005270753.1
75
20103
XM_005270697.1
XP_005270754.1
75
20103
XM_005270698.1
XP_005270755.1
75
20103
XM_006710519.1
XP_006710582.1
74
20094
XM_006710520.2
XP_006710583.1
74
20040
XM_006710521.1
XP_006710584.1
75
20103
XM_006710522.1
XP_006710585.1
75
20103
XM_006710524.2
XP_006710587.1
74
19869
XM_006710526.3
XP_006710589.1
80
19535
XM_006710528.3
XP_006710591.1
80
20096
XM_006710529.3
XP_006710592.1
80
19453
XM_006710530.3
XP_006710593.1
80
20341
XM_006710531.3
XP_006710594.1
80
20072
XM_006710532.3
XP_006710595.1
79
20000
XM_006710533.2
XP_006710596.1
79
20000
XM_006710535.3
XP_006710598.1
80
19605
XM_006710536.2
XP_006710599.1
70
13902
XM_011541141.2
XP_011539443.1
75
20049
XM_011541142.2
XP_011539444.1
74
14652
XM_017000873.1
XP_016856362.1
74
20094
XM_017000874.1
XP_016856363.1
74
20094
XM_017000875.1
XP_016856364.1
79
19425
XM_017000876.1
XP_016856365.1
79
19509
XM_017000877.1
XP_016856366.1
80
19431
XM_017000878.1
XP_016856367.1
80
19294
XM_017000879.1
XP_016856368.1
45
14865
XM_017000880.1
XP_016856369.1
69
13839
XM_017000881.1
XP_016856370.1
74
13216
XM_017000882.1
XP_016856371.1
69
13839
GenBank mRNA
AB007934.3
-
52
10199
AB029290.1
-
74
13240
AF317696.1
-
44
14815
Back To Top
More Information
Panther Classification:
Molecular Function -
intermediate filament binding protein
Gene Ontology Categories:
Function(s)
Process(es)
regulation of epithelial cell migration
Wnt signaling pathway
positive regulation of Wnt signaling pathway
regulation of microtubule-based process
wound healing
Golgi to plasma membrane protein transport
positive regulation of axon extension
regulation of focal adhesion assembly
cell-cell adhesion
actin binding
calcium ion binding
protein binding
microtubule binding
ATPase activity
poly(A) RNA binding
actin filament binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0