Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RBFOX1 SIRNA Assays ›
Gene Symbol:
RBFOX1
Gene Name:
RNA binding protein, fox-1 homolog 1
Gene Aliases:
2BP1, A2BP1, FOX-1, FOX1, HRNBP1
Chromosome Location:
Chr.16: 5239749 - 7713343 on Build GRCh38
UniGene ID:
Hs.459842
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS147742
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RBFOX1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001142333.1
NP_001135805.1
9
1569
NM_001142334.1
NP_001135806.1
7
801
NM_001308117.1
NP_001295046.1
9
906
NM_018723.3
NP_061193.2
9
1569
NM_145891.2
NP_665898.1
6
893
NM_145892.2
NP_665899.1
6
893
NM_145893.2
NP_665900.1
6
893
XM_005255386.3
XP_005255443.1
6
819
XM_005255387.3
XP_005255444.1
6
798
XM_005255388.4
XP_005255445.1
6
854
XM_005255390.3
XP_005255447.1
9
1691
XM_005255391.3
XP_005255448.1
8
701
XM_005255394.4
XP_005255451.1
6
798
XM_011522546.2
XP_011520848.1
6
798
XM_011522547.2
XP_011520849.1
6
798
XM_011522548.2
XP_011520850.1
6
798
XM_017023318.1
XP_016878807.1
10
1196
XM_017023319.1
XP_016878808.1
12
1192
XM_017023320.1
XP_016878809.1
9
1600
XM_017023321.1
XP_016878810.1
8
2662
XM_017023322.1
XP_016878811.1
8
2662
XM_017023323.1
XP_016878812.1
6
819
XM_017023324.1
XP_016878813.1
9
1600
XM_017023325.1
XP_016878814.1
7
1198
XM_017023326.1
XP_016878815.1
9
1691
XM_017023327.1
XP_016878816.1
6
819
XM_017023328.1
XP_016878817.1
9
1600
XM_017023329.1
XP_016878818.1
9
1600
XM_017023330.1
XP_016878819.1
8
2139
XM_017023331.1
XP_016878820.1
6
2605
XM_017023332.1
XP_016878821.1
6
819
XM_017023333.1
XP_016878822.1
8
701
XM_017023334.1
XP_016878823.1
6
819
XM_017023335.1
XP_016878824.1
9
1691
XM_017023336.1
XP_016878825.1
8
701
XM_017023337.1
XP_016878826.1
6
819
XM_017023338.1
XP_016878827.1
8
701
XM_017023339.1
XP_016878828.1
6
845
XM_017023340.1
XP_016878829.1
7
608
XM_017023341.1
XP_016878830.1
9
1691
XM_017023342.1
XP_016878831.1
8
701
XM_017023343.1
XP_016878832.1
6
848
GenBank mRNA
AB208826.1
-
1603
AF094849.1
-
6
894
AF107203.1
-
9
1567
AF109106.1
-
6
894
AF229057.1
-
6
896
AF448859.1
-
9
1567
AK001027.1
-
7
801
AK293951.1
-
9
906
AK299664.1
-
6
479
AL713700.1
-
8
1416
BC113691.1
-
6
687
8322743
BC143816.1
-
6
687
KJ534926.1
-
6
641
KJ535000.1
-
6
641
KJ535001.1
-
6
641
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA metabolism protein
RNA processing factor
RNA splicing factor
nucleic acid metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
regulation of alternative mRNA splicing, via spliceosome
mRNA processing
nervous system development
RNA splicing
RNA transport
nucleotide binding
RNA binding
mRNA binding
protein binding
protein C-terminus binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-9c8cf:80/100.66.75.150:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0