Search
Search

CPNE5| Gene Symbol: CPNE5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001314018.1 | NP_001300947.1 | 5, 6 | 464 | ||
| NM_001314019.1 | NP_001300948.1 | 6, 7 | 505 | |||
| NM_001314020.1 | NP_001300949.1 | 4, 5 | 402 | |||
| NM_020939.1 | NP_065990.1 | 16, 17 | 1261 | |||
| XM_005249247.2 | XP_005249304.1 | 17, 18 | 1312 | |||
| XM_011514768.1 | XP_011513070.1 | 17, 18 | 1312 | |||
| XM_011514769.1 | XP_011513071.1 | 16, 17 | 1261 | |||
| XM_011514770.1 | XP_011513072.1 | 18, 19 | 1327 | |||
| XM_011514771.2 | XP_011513073.1 | 17, 18 | 1652 | |||
| XM_011514772.1 | XP_011513074.1 | 17, 18 | 1312 | |||
| XM_011514773.2 | XP_011513075.1 | 9, 10 | 741 | |||
| XM_011514775.2 | XP_011513077.1 | 5, 6 | 455 | |||
| XM_017011139.1 | XP_016866628.1 | 15, 16 | 1182 | |||
| GenBank mRNA | AB046819.1 | - | 16, 17 | 1819 | ||
| BC053872.1 | - | 5, 6 | 464 | 5553022 | ||