Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HIF3A SIRNA Assays ›
Gene Symbol:
HIF3A
Gene Name:
hypoxia inducible factor 3 alpha subunit
Gene Aliases:
HIF-3A, HIF3-alpha-1, IPAS, MOP7, PASD7, bHLHe17
Chromosome Location:
Chr.19: 46297046 - 46343433 on Build GRCh38
UniGene ID:
Hs.420830
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS148869
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: HIF3A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_022462.4
NP_071907.4
2
151
NM_152794.3
NP_690007.1
2
101
NM_152795.3
NP_690008.2
2
69
NM_152796.2
NP_690009.1
2
151
XM_005259152.4
XP_005259209.1
3
197
XM_005259153.3
XP_005259210.2
1
14
XM_005259155.4
XP_005259212.1
3
197
XM_005259156.4
XP_005259213.1
3
197
XM_017027132.1
XP_016882621.1
3
197
XM_017027133.1
XP_016882622.1
3
197
XM_017027134.1
XP_016882623.1
2
67
XM_017027135.1
XP_016882624.1
2
101
XM_017027136.1
XP_016882625.1
3
197
XM_017027137.1
XP_016882626.1
2
101
XM_017027138.1
XP_016882627.1
2
67
XM_017027139.1
XP_016882628.1
2
137
XM_017027140.1
XP_016882629.1
2
67
XM_017027141.1
XP_016882630.1
2
109
XM_017027142.1
XP_016882631.1
2
137
GenBank mRNA
AB054067.1
-
2
32
AB118749.1
-
2
50
AB295039.1
-
2
50
AF079154.1
-
1
20
AF463492.1
-
2
64
AK021421.1
-
2
101
AK021653.1
-
2
95
AK024095.1
-
2
95
AK027725.1
-
2
50
AK297828.1
-
2
137
AK299699.1
-
1
61
AK304747.1
-
2
133
AK314680.1
-
2
101
AL541807.3
-
2
73
BC026308.1
-
2
60
4811824
BC080551.1
-
2
40
6250259
BI772109.1
-
2
126
DA559979.1
-
2
137
DA562618.1
-
2
81
DB176660.1
-
2
69
DC423791.1
-
2
151
DN989983.1
-
2
236
DR001068.1
-
2
77
Back To Top
More Information
Panther Classification:
Molecular Function -
basic helix-loop-helix transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
angiogenesis
transcription from RNA polymerase II promoter
apoptotic process
positive regulation of transcription from RNA polymerase II promoter
regulation of transcription from RNA polymerase II promoter in response to hypoxia
RNA polymerase II transcription factor activity, sequence-specific DNA binding
DNA binding
protein dimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0