Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CHD9 SIRNA Assays ›
Gene Symbol:
CHD9
Gene Name:
chromodomain helicase DNA binding protein 9
Gene Aliases:
AD013, CHD-9, CReMM, KISH2, PRIC320
Chromosome Location:
Chr.16: 53054991 - 53327502 on Build GRCh38
UniGene ID:
Hs.59159
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS149431
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CHD9
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001308319.1
NP_001295248.1
16
3963
NM_025134.4
NP_079410.4
17
4008
XM_005256168.3
XP_005256225.1
17
4050
XM_005256169.2
XP_005256226.1
17
4072
XM_005256172.3
XP_005256229.1
17
3989
XM_005256176.3
XP_005256233.1
16
2657
XM_006721281.2
XP_006721344.1
17
3989
XM_006721283.3
XP_006721346.1
17
4210
XM_011523347.2
XP_011521649.1
18
4122
XM_017023718.1
XP_016879207.1
18
4245
XM_017023719.1
XP_016879208.1
19
4196
XM_017023720.1
XP_016879209.1
18
4090
XM_017023721.1
XP_016879210.1
18
4246
XM_017023722.1
XP_016879211.1
18
4246
XM_017023723.1
XP_016879212.1
18
4246
XM_017023724.1
XP_016879213.1
18
4072
XM_017023725.1
XP_016879214.1
18
4072
XM_017023726.1
XP_016879215.1
16
3279
XM_017023727.1
XP_016879216.1
16
2657
XM_017023728.1
XP_016879217.1
18
4106
GenBank mRNA
AB002306.3
-
16
3431
AK022582.1
-
6
1146
AK299045.1
-
16
2706
AY243500.1
-
16
3886
AY647157.1
-
17
4008
BC140815.1
-
16
3840
9021683
BC144622.1
-
16
3840
9053153
DQ059482.1
-
7
1336
DQ333316.1
-
17
4102
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: XM_008772343.
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
covalent chromatin modification
cellular lipid metabolic process
DNA binding
helicase activity
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0