Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CDK5RAP3 SIRNA Assays ›
          Gene Symbol:
          CDK5RAP3
          Gene Name:
          CDK5 regulatory subunit associated protein 3
          Gene Aliases:
          C53, HSF-27, IC53, LZAP, MST016, OK/SW-cl.114, PP1553
          Chromosome Location:
          Chr.17: 47967904 - 47981786 on Build GRCh38
          UniGene ID:
          Hs.20157
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS149443
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CDK5RAP3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001278197.1 NP_001265126.1 11 1211
          NM_001278198.1 NP_001265127.1 9 2137
          NM_001278216.1 NP_001265145.1 9 2137
          NM_001278217.1 NP_001265146.1 10 1922
          NM_176096.2 NP_788276.1 11 1212
          XM_011525297.1 XP_011523599.1 12 1152
          XM_017025164.1 XP_016880653.1 10 1871
          XM_017025165.1 XP_016880654.1 9 2131
          XM_017025166.1 XP_016880655.1 11 1161
          GenBank mRNA AB062433.1 - 10 1026
          AF110322.1 - 10 1753
          AF217982.2 - 9 1989
          AF343090.1 - 11 1056
          AK000260.1 - 5 1466
          AK023722.1 - 11 1078
          AK223387.1 - 11 1127
          AK225274.1 - 9 1966
          AK300643.1 - 11 1205
          BC009957.2 - 10 1026 4130677
          BC072435.1 - 11 1038 5751801

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of cyclin-dependent protein serine/threonine kinase activity
          negative regulation of protein phosphorylation
          mitotic G2 DNA damage checkpoint
          brain development
          cell proliferation
          regulation of phosphatase activity
          apoptotic nuclear changes
          endoplasmic reticulum unfolded protein response
          positive regulation of protein ubiquitination
          negative regulation of NF-kappaB transcription factor activity
          negative regulation of MAP kinase activity
          negative regulation of protein kinase activity by regulation of protein phosphorylation
          mitotic G2/M transition checkpoint
          regulation of neuron differentiation
          positive regulation of transcription from RNA polymerase II promoter
          protein ufmylation
          negative regulation of protein serine/threonine kinase activity
          positive regulation of protein localization to nucleus
          positive regulation of signal transduction by p53 class mediator
          negative regulation of cellular protein catabolic process
          positive regulation of protein ubiquitination involved in ubiquitin-dependent protein catabolic process
          protein binding
          protein kinase binding
          cyclin binding
          protein complex binding
          ubiquitin-like protein ligase binding
          mitogen-activated protein kinase binding
          NF-kappaB binding
          MDM2/MDM4 family protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0