Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ASCC2 SIRNA Assays ›
Gene Symbol:
ASCC2
Gene Name:
activating signal cointegrator 1 complex subunit 2
Gene Aliases:
ASC1p100, p100
Chromosome Location:
Chr.22: 29788608 - 29838304 on Build GRCh38
UniGene ID:
Hs.731754
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS149845
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ASCC2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242906.1
NP_001229835.1
12
1114
NM_032204.4
NP_115580.2
13
1342
XM_005261775.2
XP_005261832.1
14
1382
XM_011530442.2
XP_011528744.1
14
1382
XM_011530443.2
XP_011528745.1
15
1448
XM_011530444.2
XP_011528746.1
15
1495
XM_011530445.2
XP_011528747.1
14
1429
XM_011530446.2
XP_011528748.1
13
1312
XM_011530448.2
XP_011528750.1
13
1215
XM_011530449.2
XP_011528751.1
12
1153
XM_011530450.2
XP_011528752.1
15
1498
XM_011530451.2
XP_011528753.1
16
1572
XM_011530452.2
XP_011528754.1
15
1545
XM_011530453.2
XP_011528755.1
14
1400
XM_011530454.2
XP_011528756.1
13
1284
XM_011530455.2
XP_011528757.1
12
1213
XM_017028991.1
XP_016884480.1
13
3520
XM_017028992.1
XP_016884481.1
13
3520
XM_017028993.1
XP_016884482.1
12
3445
XM_017028994.1
XP_016884483.1
12
3445
XM_017028995.1
XP_016884484.1
12
3361
XM_017028996.1
XP_016884485.1
14
2322
XM_017028997.1
XP_016884486.1
12
3361
XM_017028998.1
XP_016884487.1
14
2322
XM_017028999.1
XP_016884488.1
15
1448
XM_017029000.1
XP_016884489.1
14
1429
XM_017029001.1
XP_016884490.1
13
1215
XM_017029002.1
XP_016884491.1
12
1153
XM_017029003.1
XP_016884492.1
14
1289
XM_017029004.1
XP_016884493.1
11
1078
XM_017029005.1
XP_016884494.1
15
1498
XM_017029006.1
XP_016884495.1
16
1572
XM_017029007.1
XP_016884496.1
14
1400
XM_017029008.1
XP_016884497.1
13
1284
XM_017029009.1
XP_016884498.1
12
1213
GenBank mRNA
AK022886.1
-
15
1460
AK025241.1
-
5
371
AK222723.1
-
5
371
AK223033.1
-
5
370
AK303257.1
-
12
1084
AL096741.1
-
1
32
AY013289.1
-
12
1219
BC025368.2
-
13
1303
3955747
Back To Top
More Information
Gene Ontology Categories:
Function(s)
DNA dealkylation involved in DNA repair
transcription, DNA-templated
regulation of transcription, DNA-templated
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0