Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other IL17RC SIRNA Assays ›
          Gene Symbol:
          IL17RC
          Gene Name:
          interleukin 17 receptor C
          Gene Aliases:
          CANDF9, IL17-RL, IL17RL
          Chromosome Location:
          Chr.3: 9917074 - 9933627 on Build GRCh38
          UniGene ID:
          Hs.129959
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS150138
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price 459.00
          Your Price
          Online offer:
          459.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: IL17RC
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001203265.1 NP_001190194.1 15, 16 1631
          NM_032732.5 NP_116121.2 16, 17 1682
          NM_153460.3 NP_703190.1 17, 18 1727
          NM_153461.3 NP_703191.1 17, 18 1916
          NR_037807.1 15, 16 1893
          XM_017007326.1 XP_016862815.1 17, 18 1919
          XM_017007328.1 XP_016862817.1 17, 18 1880
          XM_017007330.1 XP_016862819.1 17, 18 1877
          XM_017007331.1 XP_016862820.1 16, 17 1874
          XM_017007332.1 XP_016862821.1 16, 17 1871
          XM_017007333.1 XP_016862822.1 16, 17 1865
          XM_017007334.1 XP_016862823.1 16, 17 1844
          XM_017007335.1 XP_016862824.1 16, 17 1832
          XM_017007337.1 XP_016862826.1 15, 16 1799
          XM_017007340.1 XP_016862829.1 17, 18 1707
          XM_017007341.1 XP_016862830.1 16, 17 1662
          XM_017007342.1 XP_016862831.1 16, 17 1631
          XM_017007343.1 XP_016862832.1 16, 17 1628
          XM_017007345.1 XP_016862834.1 16, 17 1623
          XM_017007347.1 XP_016862836.1 15, 16 1586
          XM_017007348.1 XP_016862837.1 15, 16 1569
          GenBank mRNA AF458065.1 - 17, 18 1682
          AK075534.1 - 14, 15 1798
          AK092907.1 - 17, 18 1917
          AK096824.1 - 14, 15 1645
          AK315024.1 - 15, 16 1870
          AK315053.1 - 16, 17 1627
          AY358840.1 - 16, 17 1644
          AY359098.1 - 16, 17 1672
          EF676032.1 - 15, 16 1542

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          cytokine-mediated signaling pathway
          defense response to fungus
          granulocyte chemotaxis
          positive regulation of cytokine production involved in inflammatory response
          positive regulation of interleukin-6 secretion
          receptor binding
          interleukin-17 receptor activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0