Search
Search

LINC00313
LOC102724354| Gene Symbol: LINC00313 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026863.1 | 3 | 270 | |||
| GenBank mRNA | AF426261.1 | - | 3 | 270 | ||
| BC107109.2 | - | 3 | 201 | |||
| Gene Symbol: LOC102724354 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136540.1 | 4 | 231 | |||
| GenBank mRNA | AW206028.1 | - | 2 | 305 | ||
| HY162931.1 | - | 3 | 189 | |||
| HY359299.1 | - | 2 | 440 | |||