Search
Search

FCRLB| Gene Symbol: FCRLB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002901.3 | NP_001002901.1 | 8 | 1398 | ||
| NM_001288829.1 | NP_001275758.1 | 6 | 1039 | |||
| NM_001288830.1 | NP_001275759.1 | 5 | 1018 | |||
| NM_001288831.1 | NP_001275760.1 | 6 | 1080 | |||
| NM_001288832.1 | NP_001275761.1 | 5 | 1059 | |||
| NM_001320241.1 | NP_001307170.1 | 7 | 1212 | |||
| GenBank mRNA | AK055614.1 | - | 2 | 1392 | ||
| AK290733.1 | - | 7 | 1212 | |||
| AY670683.1 | - | 6 | 1179 | |||
| AY670684.1 | - | 6 | 1034 | |||
| AY670685.1 | - | 5 | 1013 | |||
| AY670686.1 | - | 6 | 1075 | |||
| AY670687.1 | - | 5 | 1054 | |||
| AY786314.1 | - | 6 | 1164 | |||
| BC038564.2 | - | 3 | 764 | |||
| BC067080.1 | - | 2 | 1381 | |||
| DQ021957.1 | - | 8 | 1398 | |||