Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MIB2 SIRNA Assays ›
          Gene Symbol:
          MIB2
          Gene Name:
          mindbomb E3 ubiquitin protein ligase 2
          Gene Aliases:
          ZZANK1, ZZZ5
          Chromosome Location:
          Chr.1: 1615188 - 1630610 on Build GRCh38
          UniGene ID:
          Hs.135805
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS152026
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MIB2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001170686.1 NP_001164157.1 14 2140
          NM_001170687.1 NP_001164158.1 14 2110
          NM_001170688.1 NP_001164159.1 13 1957
          NM_001170689.1 NP_001164160.1 13 1932
          NM_080875.2 NP_543151.2 14 2152
          NR_033183.1 14 1935
          XM_006710372.1 XP_006710435.1 13 2016
          XM_011540731.2 XP_011539033.1 15 2889
          XM_011540736.2 XP_011539038.1 15 2889
          XM_011540737.2 XP_011539039.1 14 2826
          XM_011540741.2 XP_011539043.1 13 2721
          XM_011540742.2 XP_011539044.1 13 2718
          XM_017000349.1 XP_016855838.1 15 2889
          XM_017000350.1 XP_016855839.1 15 2877
          XM_017000351.1 XP_016855840.1 15 2871
          XM_017000352.1 XP_016855841.1 15 2847
          XM_017000353.1 XP_016855842.1 14 2826
          XM_017000354.1 XP_016855843.1 15 2889
          XM_017000355.1 XP_016855844.1 13 2721
          XM_017000356.1 XP_016855845.1 13 2718
          XM_017000357.1 XP_016855846.1 14 2694
          XM_017000358.1 XP_016855847.1 15 2187
          XM_017000359.1 XP_016855848.1 15 2175
          XM_017000360.1 XP_016855849.1 15 2134
          XM_017000361.1 XP_016855850.1 14 2124
          XM_017000362.1 XP_016855851.1 15 2227
          XM_017000363.1 XP_016855852.1 15 2190
          XM_017000364.1 XP_016855853.1 13 1929
          XM_017000365.1 XP_016855854.1 15 1917
          XM_017000366.1 XP_016855855.1 14 1854
          GenBank mRNA AB074480.1 - 14 2063
          AB076691.1 - 14 2021
          AB076692.1 - 13 1868
          AB076693.1 - 14 2051
          AB097000.1 - 14 1986
          AK091610.1 - 8 956
          AK095914.1 - 8 958
          AK096295.1 - 9 1063
          AK097106.1 - 10 1382
          AK098785.1 - 14 2021
          AK122751.1 - 14 1854
          AK122863.1 - 14 2053
          AK128167.1 - 13 1892
          AK128553.1 - 13 3066
          AK293642.1 - 13 2132
          AK302757.1 - 14 2104
          AK309362.1 - 12 2022
          AL834527.1 - 9 1184
          BC016490.1 - 14 1989
          BC037542.1 - 14 2032 5168033

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein polyubiquitination
          Notch signaling pathway
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          actin binding
          ubiquitin-protein transferase activity
          signal transducer activity
          protein binding
          zinc ion binding
          ligase activity
          ubiquitin protein ligase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-9rjdb:80/100.66.79.224:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline