Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MIB2 SIRNA Assays ›
Gene Symbol:
MIB2
Gene Name:
mindbomb E3 ubiquitin protein ligase 2
Gene Aliases:
ZZANK1, ZZZ5
Chromosome Location:
Chr.1: 1615188 - 1630610 on Build GRCh38
UniGene ID:
Hs.135805
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS152027
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MIB2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001170686.1
NP_001164157.1
16
2448
NM_001170687.1
NP_001164158.1
16
2418
NM_001170688.1
NP_001164159.1
15
2265
NM_001170689.1
NP_001164160.1
15
2240
NM_080875.2
NP_543151.2
16
2460
NR_033183.1
16
2243
XM_006710372.1
XP_006710435.1
15
2324
XM_011540731.2
XP_011539033.1
17
3197
XM_011540736.2
XP_011539038.1
17
3197
XM_011540737.2
XP_011539039.1
16
3134
XM_011540741.2
XP_011539043.1
15
3029
XM_011540742.2
XP_011539044.1
15
3026
XM_017000349.1
XP_016855838.1
17
3197
XM_017000350.1
XP_016855839.1
17
3185
XM_017000351.1
XP_016855840.1
17
3179
XM_017000352.1
XP_016855841.1
17
3155
XM_017000353.1
XP_016855842.1
16
3134
XM_017000354.1
XP_016855843.1
17
3197
XM_017000355.1
XP_016855844.1
15
3029
XM_017000356.1
XP_016855845.1
15
3026
XM_017000357.1
XP_016855846.1
16
3002
XM_017000358.1
XP_016855847.1
17
2495
XM_017000359.1
XP_016855848.1
17
2483
XM_017000360.1
XP_016855849.1
17
2442
XM_017000361.1
XP_016855850.1
16
2432
XM_017000362.1
XP_016855851.1
17
2535
XM_017000363.1
XP_016855852.1
17
2498
XM_017000364.1
XP_016855853.1
15
2237
XM_017000365.1
XP_016855854.1
17
2225
XM_017000366.1
XP_016855855.1
16
2162
GenBank mRNA
AB074480.1
-
16
2371
AB076691.1
-
16
2329
AB076692.1
-
15
2176
AB076693.1
-
16
2359
AB097000.1
-
16
2294
AK091610.1
-
10
1264
AK095914.1
-
10
1266
AK096295.1
-
11
1371
AK097106.1
-
12
1690
AK098785.1
-
16
2329
AK122751.1
-
16
2162
AK122863.1
-
15
2454
AK128167.1
-
15
2200
AK128553.1
-
15
3374
AK293642.1
-
15
2440
AK302757.1
-
16
2412
AK309362.1
-
13
2419
AL834527.1
-
11
1492
BC016490.1
-
16
2297
BC037542.1
-
16
2340
5168033
Back To Top
More Information
Panther Classification:
Molecular Function -
ubiquitin-protein ligase
Gene Ontology Categories:
Function(s)
Process(es)
protein polyubiquitination
Notch signaling pathway
positive regulation of I-kappaB kinase/NF-kappaB signaling
actin binding
ubiquitin-protein transferase activity
signal transducer activity
protein binding
zinc ion binding
ligase activity
ubiquitin protein ligase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-qfdf2:80/100.66.76.55:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline