Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SLC2A14 SIRNA Assays ›
Gene Symbol:
SLC2A14
Gene Name:
solute carrier family 2 member 14
Gene Aliases:
GLUT14, SLC2A3P3
Chromosome Location:
Chr.12: 7812512 - 7891196 on Build GRCh38
UniGene ID:
Hs.655169
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS152072
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SLC2A14
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001286233.1
NP_001273162.1
11
1655
NM_001286234.1
NP_001273163.1
6
901
NM_001286235.1
NP_001273164.1
8
1199
NM_001286236.1
NP_001273165.1
5
740
NM_001286237.1
NP_001273166.1
5
794
NM_153449.3
NP_703150.1
7
937
XM_005253315.3
XP_005253372.1
6
735
XM_005253317.4
XP_005253374.1
6
1254
XM_011520562.1
XP_011518864.1
9
1089
XM_011520563.2
XP_011518865.1
6
749
XM_011520564.2
XP_011518866.1
5
558
XM_011520565.2
XP_011518867.1
4
587
XM_017018841.1
XP_016874330.1
7
834
XM_017018842.1
XP_016874331.1
10
1405
XM_017018843.1
XP_016874332.1
10
1729
XM_017018844.1
XP_016874333.1
7
784
XM_017018845.1
XP_016874334.1
7
1326
XM_017018846.1
XP_016874335.1
7
910
XM_017018847.1
XP_016874336.1
6
867
GenBank mRNA
AF481878.1
-
6
720
AF481879.1
-
7
797
AK122783.1
-
8
987
AK126026.1
-
11
1437
AK299037.1
-
5
746
AK302881.1
-
5
553
AK316238.1
-
5
746
AL832448.1
-
4
561
AY357941.1
-
6
886
BC060766.1
-
6
748
5297510
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
multicellular organism development
spermatogenesis
cell differentiation
glucose transmembrane transport
glucose transmembrane transporter activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-tv4sr:80/100.66.76.55:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline