Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PLB1 SIRNA Assays ›
Gene Symbol:
PLB1
Gene Name:
phospholipase B1
Gene Aliases:
PLB, PLB/LIP, hPLB
Chromosome Location:
Chr.2: 28488579 - 28644135 on Build GRCh38
UniGene ID:
Hs.444933
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS152593
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PLB1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001170585.1
NP_001164056.1
23
1591
NM_153021.4
NP_694566.4
23
1558
XM_011532579.2
XP_011530881.1
25
1865
XM_011532581.2
XP_011530883.1
25
1828
XM_011532584.2
XP_011530886.1
25
1865
XM_011532588.2
XP_011530890.1
25
1867
XM_011532589.2
XP_011530891.1
25
1862
XM_011532590.2
XP_011530892.1
25
1867
XM_011532591.2
XP_011530893.1
25
1864
XM_011532593.2
XP_011530895.1
21
1440
XM_011532601.2
XP_011530903.1
11
853
XM_011532602.2
XP_011530904.1
10
825
XM_011532603.2
XP_011530905.1
10
902
XM_011532606.2
XP_011530908.1
25
1887
XM_011532607.2
XP_011530909.1
25
1887
XM_011532608.2
XP_011530910.1
25
1885
XM_011532609.2
XP_011530911.1
10
823
XM_011532610.2
XP_011530912.1
2
1009
XM_017003432.1
XP_016858921.1
24
1834
XM_017003433.1
XP_016858922.1
24
1800
XM_017003434.1
XP_016858923.1
25
1869
GenBank mRNA
AK055428.1
-
4
241
BC065041.1
-
10
644
BC150633.1
-
23
1558
9057003
BC153864.1
-
23
1543
Back To Top
More Information
Panther Classification:
Molecular Function -
hydrolase
lipase
phospholipase
metabolite interconversion enzyme
Gene Ontology Categories:
Function(s)
Process(es)
retinoid metabolic process
lipid catabolic process
phosphatidylcholine acyl-chain remodeling
retinol metabolic process
positive regulation of acrosome reaction
lysophospholipase activity
phospholipase A2 activity
retinyl-palmitate esterase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0