Search
Search

SIGLEC17P| Gene Symbol: SIGLEC17P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_047529.2 | 2 | 219 | |||
| NR_047530.2 | 2 | 219 | ||||
| NR_047531.2 | 2 | 219 | ||||
| GenBank mRNA | AF150143.1 | - | 2 | 150 | ||
| AF161341.1 | - | 2 | 150 | |||
| AK301336.1 | - | 2 | 149 | |||
| BC041072.1 | - | 2 | 219 | |||
| BC050999.1 | - | 2 | 219 | |||
| DC340819.1 | - | 2 | 149 | |||