Search
Search

| Gene Symbol: HLA-F-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026972.1 | 4 | 1022 | |||
| GenBank mRNA | AK092748.1 | - | 4 | 1022 | ||
| AL832418.1 | - | 5 | 841 | |||
| Gene Symbol: HLA-F | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011514564.1 | XP_011512866.1 | 6 | 1087 | ||
| XM_017010813.1 | XP_016866302.1 | 8 | 1242 | |||
| GenBank mRNA | AK096962.1 | - | 6 | 1077 | ||
| AK301488.1 | - | 8 | 1346 | |||