Search
Search

| Gene Symbol: CCDC103 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001258395.1 | NP_001245324.1 | 2 | 176 | ||
| NM_001258396.1 | NP_001245325.1 | 2 | 190 | |||
| NM_001258397.1 | NP_001245326.1 | 2 | 206 | |||
| NM_001258398.1 | NP_001245327.1 | 2 | 190 | |||
| NM_001258399.1 | NP_001245328.1 | 2 | 206 | |||
| NM_213607.2 | NP_998772.1 | 2 | 206 | |||
| GenBank mRNA | AK091530.1 | - | 2 | 133 | ||
| AK289760.1 | - | 2 | 136 | |||
| BC041060.1 | - | 2 | 132 | 5455678 | ||
| DA185416.1 | - | 2 | 176 | |||
| DA749894.1 | - | 2 | 179 | |||
| DB085134.1 | - | 2 | 186 | |||
| Gene Symbol: FAM187A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK023156.1 | - | 2 | 144 | ||