Search
Search

| Gene Symbol: FARSB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017003110.1 | XP_016858599.1 | 3 | 677 | ||
| GenBank mRNA | AK001025.1 | - | 3 | 666 | ||
| Gene Symbol: TRIM61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK296100.1 | - | 1 | 1031 | ||