Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other GOLGA6L22 SIRNA Assays ›
          Gene Symbol:
          GOLGA6L22 GOLGA6L1 GOLGA6L6 LOC102723623 MATR3 PPARA PSMD5-AS1 TCAM1P
          Gene Name:
          golgin A6 family-like 22
          golgin A6 family-like 1
          golgin A6 family-like 6
          golgin subfamily A member 6-like protein 6
          matrin 3
          peroxisome proliferator activated receptor alpha
          PSMD5 antisense RNA 1 (head to head)
          testicular cell adhesion molecule 1, pseudogene
          Gene Aliases:
          -
          -
          -
          -
          ALS21, MPD2, VCPDM
          NR1C1, PPAR, PPARalpha, hPPAR
          -
          TCAM1
          Chromosome Location:
          Chr.15: 22458903 - 22468026 on Build GRCh38
          Chromosome Location:
          Chr.15: 23127066 - 23136839 on Build GRCh38
          Chromosome Location:
          Chr.15: 20531856 - 20541800 on Build GRCh38
          Chromosome Location:
          Chr.15: 23324535 - 23334306 on Build GRCh38
          Chromosome Location:
          Chr.5: 139273752 - 139331677 on Build GRCh38
          Chromosome Location:
          Chr.22: 46150534 - 46243756 on Build GRCh38
          Chromosome Location:
          Chr.9: 120843042 - 120854373 on Build GRCh38
          Chromosome Location:
          Chr.17: 63857016 - 63864379 on Build GRCh38
          UniGene ID:
          -
          Hs.531569
          Hs.569472
          -
          Hs.268939
          Hs.103110
          Hs.594170
          Hs.629177
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          -
          siRNA ID HSS162870
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: GOLGA6L22
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001271664.2 NP_001258593.2 10 3382
          Gene Symbol: GOLGA6L1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001001413.3 NP_001001413.3 9 3398
          Gene Symbol: GOLGA6L6
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001145004.2 NP_001138476.2 9 3589
          GenBank mRNA AK093450.1 - 2 1554
          Gene Symbol: LOC102723623
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_006720795.3 XP_006720858.1 9 3417
          XM_017022778.1 XP_016878267.1 10 3490
          Gene Symbol: MATR3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA BX647153.1 - 1 1983
          Gene Symbol: PPARA
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001001928.2 NP_001001928.1 8 2204
          NM_005036.4 NP_005027.2 9 2288
          XM_005261655.3 XP_005261712.1 9 2581
          XM_005261656.3 XP_005261713.1 10 2745
          XM_006724269.3 XP_006724332.1 9 2347
          XM_006724270.3 XP_006724333.1 8 2253
          XM_011530239.2 XP_011528541.1 10 2437
          XM_011530240.2 XP_011528542.1 10 2440
          XM_011530241.2 XP_011528543.1 9 2345
          XM_011530242.2 XP_011528544.1 8 2167
          XM_011530243.2 XP_011528545.1 7 2408
          XM_011530244.2 XP_011528546.1 10 2444
          XM_011530245.2 XP_011528547.1 8 2260
          XM_017028839.1 XP_016884328.1 11 2619
          GenBank mRNA AF018254.1 - 1 111
          Gene Symbol: PSMD5-AS1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AK123991.1 - 1 922
          Gene Symbol: TCAM1P
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA LC005099.1 - 1 19

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          actin or actin-binding cytoskeletal protein C4 zinc finger nuclear receptor DNA-binding transcription factor zinc finger transcription factor gene-specific transcriptional regulator

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          response to hypoxia
          transcription initiation from RNA polymerase II promoter
          lipid metabolic process
          fatty acid metabolic process
          heart development
          epidermis development
          negative regulation of macrophage derived foam cell differentiation
          negative regulation of receptor biosynthetic process
          negative regulation of cholesterol storage
          negative regulation of sequestering of triglyceride
          fatty acid transport
          intracellular receptor signaling pathway
          positive regulation of fatty acid beta-oxidation
          negative regulation of protein binding
          negative regulation of appetite
          response to insulin
          circadian regulation of gene expression
          behavioral response to nicotine
          wound healing
          lipoprotein metabolic process
          regulation of circadian rhythm
          steroid hormone mediated signaling pathway
          cellular lipid metabolic process
          positive regulation of gluconeogenesis
          negative regulation of blood pressure
          negative regulation of glycolytic process
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of fatty acid oxidation
          negative regulation of inflammatory response
          enamel mineralization
          regulation of glycolytic process by positive regulation of transcription from RNA polymerase II promoter
          regulation of cellular ketone metabolic process by positive regulation of transcription from RNA polymerase II promoter
          regulation of lipid transport by positive regulation of transcription from RNA polymerase II promoter
          negative regulation of neuron death
          negative regulation of pri-miRNA transcription from RNA polymerase II promoter
          negative regulation of leukocyte cell-cell adhesion
          negative regulation of transcription regulatory region DNA binding
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
          RNA polymerase II repressing transcription factor binding
          transcriptional activator activity, RNA polymerase II transcription factor binding
          transcription coactivator binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          steroid hormone receptor activity
          RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
          protein binding
          transcription factor binding
          drug binding
          zinc ion binding
          lipid binding
          phosphatase binding
          protein domain specific binding
          ubiquitin conjugating enzyme binding
          protein complex binding
          sequence-specific DNA binding
          NFAT protein binding
          MDM2/MDM4 family protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0