Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NPIPB9 SIRNA Assays ›
          Gene Symbol:
          NPIPB9 NPIPB6 NPIPB8
          Gene Name:
          nuclear pore complex interacting protein family member B9
          nuclear pore complex interacting protein family member B6
          nuclear pore complex interacting protein family member B8
          Gene Aliases:
          -
          NPIPB
          -
          Chromosome Location:
          Chr.16: 28735500 - 28772911 on Build GRCh38
          Chromosome Location:
          Chr.16: 28342517 - 28364971 on Build GRCh38
          Chromosome Location:
          Chr.16: 28638065 - 28658693 on Build GRCh38
          UniGene ID:
          Hs.710214
          Hs.726467
          -
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          siRNA ID HSS162907
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NPIPB9
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001287250.1 NP_001274179.1 3 387
          NM_001287251.1 NP_001274180.1 4 473
          XM_003118719.7 XP_003118767.2 4 948
          XM_011545690.2 XP_011543992.1 5 1037
          XM_011545693.2 XP_011543995.1 5 1036
          XM_017022814.1 XP_016878303.1 5 1037
          XM_017022815.1 XP_016878304.1 5 557
          XM_017022816.1 XP_016878305.1 5 1037
          XM_017022817.1 XP_016878306.1 4 949
          XM_017022818.1 XP_016878307.1 4 2274
          XM_017022819.1 XP_016878308.1 5 964
          XM_017022820.1 XP_016878309.1 5 681
          XM_017022821.1 XP_016878310.1 4 1547
          XM_017022822.1 XP_016878311.1 5 2550
          XM_017022823.1 XP_016878312.1 3 2186
          XM_017022824.1 XP_016878313.1 5 963
          XM_017022825.1 XP_016878314.1 5 681
          XM_017022826.1 XP_016878315.1 4 498
          XM_017022828.1 XP_016878317.1 5 1036
          GenBank mRNA BC171825.1 - 4 1123
          Gene Symbol: NPIPB6
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001282524.1 NP_001269453.1 3 387
          XM_005255741.3 XP_005255798.1 4 1199
          XM_005255742.4 XP_005255799.1 4 945
          XM_011545929.2 XP_011544231.1 5 1019
          XM_011545931.2 XP_011544233.1 4 652
          XM_017023628.1 XP_016879117.1 4 402
          XM_017023629.1 XP_016879118.1 4 945
          XM_017023630.1 XP_016879119.1 5 672
          XM_017023631.1 XP_016879120.1 3 1016
          XM_017023632.1 XP_016879121.1 4 402
          GenBank mRNA BC144439.1 - 3 348
          BC157836.1 - 3 1037 9057055
          KF175222.1 - 3 349
          Gene Symbol: NPIPB8
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001310136.1 NP_001297065.1 3 349
          XM_017023622.1 XP_016879111.1 4 473
          XM_017023623.1 XP_016879112.1 4 473
          XM_017023624.1 XP_016879113.1 4 473
          XM_017023625.1 XP_016879114.1 4 453
          XM_017023626.1 XP_016879115.1 5 1092
          XM_017023627.1 XP_016879116.1 4 473

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0