Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other QRSL1 SIRNA Assays ›
          Gene Symbol:
          QRSL1 MAATS1 METTL15 PCMTD2 ZC3H12B ZNF337-AS1
          Gene Name:
          glutaminyl-tRNA synthase (glutamine-hydrolyzing)-like 1
          MYCBP associated and testis expressed 1
          methyltransferase like 15
          protein-L-isoaspartate (D-aspartate) O-methyltransferase domain containing 2
          zinc finger CCCH-type containing 12B
          ZNF337 antisense RNA 1
          Gene Aliases:
          GatA
          AAT1, AAT1alpha, C3orf15, CaM-IP2, SPATA26
          METT5D1
          C20orf36
          CXorf32, MCPIP2
          -
          Chromosome Location:
          Chr.6: 106629566 - 106668417 on Build GRCh38
          Chromosome Location:
          Chr.3: 119703022 - 119767102 on Build GRCh38
          Chromosome Location:
          Chr.11: 28108251 - 28527041 on Build GRCh38
          Chromosome Location:
          Chr.20: 64255695 - 64276226 on Build GRCh38
          Chromosome Location:
          Chr.X: 65034815 - 65507887 on Build GRCh38
          Chromosome Location:
          Chr.20: 25624045 - 25678074 on Build GRCh38
          UniGene ID:
          Hs.406917
          Hs.341906
          Hs.243326
          Hs.744845
          Hs.21249
          Hs.633621
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          siRNA ID HSS163704
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: QRSL1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_018292.4 NP_060762.3 11 1935
          XM_011535924.2 XP_011534226.1 12 2029
          GenBank mRNA AK001851.1 - 11 1917
          AK023509.1 - 11 1869
          AK225262.1 - 11 1923
          AL136679.1 - 11 1923
          BC006084.1 - 11 1895 3605968
          BC014389.2 - 11 1920 3504569
          Gene Symbol: MAATS1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AL133066.1 - 4 1801
          Gene Symbol: METTL15
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AJ131606.1 - 1 255
          Gene Symbol: PCMTD2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA BC032332.1 - 7 1582 5169056
          Gene Symbol: ZC3H12B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001010888.3 NP_001010888.3 5 3077
          XM_011530939.2 XP_011529241.1 6 3322
          XM_017029478.1 XP_016884967.1 11 4588
          XM_017029479.1 XP_016884968.1 13 7881
          XM_017029480.1 XP_016884969.1 11 4622
          XM_017029481.1 XP_016884970.1 11 4617
          XM_017029482.1 XP_016884971.1 10 4527
          XM_017029483.1 XP_016884972.1 9 4323
          XM_017029484.1 XP_016884973.1 9 3955
          GenBank mRNA BC140767.1 - 5 3077 9021635
          BC171781.1 - 5 3077 9054435
          BX647241.1 - 7 2827
          Gene Symbol: ZNF337-AS1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_126466.1 3 261
          NR_126467.1 4 503
          GenBank mRNA AL529206.3 - 4 503
          BC034569.1 - 4 392
          CN421644.1 - 3 253

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          ligase methyltransferase endoribonuclease

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of protein stability
          mitochondrial translation
          glutaminyl-tRNAGln biosynthesis via transamidation
          nucleic acid phosphodiester bond hydrolysis
          ATP binding
          glutaminyl-tRNA synthase (glutamine-hydrolyzing) activity
          endonuclease activity
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0