Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other POTEF SIRNA Assays ›
          Gene Symbol:
          POTEF POTEE POTEI POTEJ POTEKP
          Gene Name:
          POTE ankyrin domain family member F
          POTE ankyrin domain family member E
          POTE ankyrin domain family member I
          POTE ankyrin domain family member J
          POTE ankyrin domain family member K, pseudogene
          Gene Aliases:
          A26C1B, ACTB, POTE2alpha, POTEACTIN
          A26C1, A26C1A, CT104.2, POTE-2, POTE2, POTE2gamma
          POTE2beta
          POTE2beta
          ACT, ACTBL3, POTE2delta, POTEK
          Chromosome Location:
          Chr.2: 130073535 - 130129222 on Build GRCh38
          Chromosome Location:
          Chr.2: 131209580 - 131265254 on Build GRCh38
          Chromosome Location:
          Chr.2: 130462245 - 131626428 on Build GRCh38
          Chromosome Location:
          Chr.2: 130611077 - 130658448 on Build GRCh38
          Chromosome Location:
          Chr.2: 131592093 - 131609401 on Build GRCh38
          UniGene ID:
          Hs.580547
          Hs.686146
          Hs.558782
          Hs.558829
          Hs.631267
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          siRNA ID HSS163962
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: POTEF
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001099771.2 NP_001093241.1 17 2494
          XM_017004833.1 XP_016860322.1 15 2269
          XM_017004834.1 XP_016860323.1 10 991
          XM_017004835.1 XP_016860324.1 8 928
          GenBank mRNA EF523384.1 - 17 2496
          Gene Symbol: POTEE
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001083538.1 NP_001077007.1 15 2146
          XM_011511216.2 XP_011509518.1 11 1870
          XM_017004159.1 XP_016859648.1 11 1901
          XM_017004160.1 XP_016859649.1 7 869
          XM_017004161.1 XP_016859650.1 17 2583
          GenBank mRNA AY462868.1 - 12 1985
          AY462871.1 - 15 2146
          AY462872.1 - 14 2104
          AY462873.1 - 13 2030
          Gene Symbol: POTEI
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001277406.1 NP_001264335.1 15 2094
          XM_011511670.2 XP_011509972.1 15 2266
          XM_017004732.1 XP_016860221.1 16 2245
          XM_017004733.1 XP_016860222.1 10 991
          XM_017004734.1 XP_016860223.1 8 928
          Gene Symbol: POTEJ
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001277083.1 NP_001264012.1 15 1983
          XM_011511683.2 XP_011509985.1 15 2155
          XM_017004741.1 XP_016860230.1 16 2128
          XM_017004742.1 XP_016860231.1 10 991
          XM_017004743.1 XP_016860232.1 8 923
          Gene Symbol: POTEKP
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AY014272.1 - 1 18

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          actin and actin related protein actin or actin-binding cytoskeletal protein cytoskeletal protein membrane traffic protein

          Gene Ontology Categories:

          Function(s) Process(es)

          retina homeostasis
          substantia nigra development
          molecular_function

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0