Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LINC00999 SIRNA Assays ›
          Gene Symbol:
          LINC00999 LINC00265 LINC01000 LINC01001 LINC01002 LINC01061 LOC100132062 LOC100132217 LOC100132287 LOC100133331 LOC107986805 LOC401357 LOC729737
          Gene Name:
          long intergenic non-protein coding RNA 999
          long intergenic non-protein coding RNA 265
          long intergenic non-protein coding RNA 1000
          long intergenic non-protein coding RNA 1001
          long intergenic non-protein coding RNA 1002
          long intergenic non-protein coding RNA 1061
          uncharacterized LOC100132062
          uncharacterized LOC100132217
          uncharacterized LOC100132287
          uncharacterized LOC100133331
          putative uncharacterized protein FLJ44672
          uncharacterized LOC401357
          uncharacterized LOC729737
          Gene Aliases:
          -
          NCRNA00265
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          Chromosome Location:
          Chr.10: 38428146 - 38452153 on Build GRCh38
          Chromosome Location:
          Chr.7: 39733568 - 39794623 on Build GRCh38
          Chromosome Location:
          Chr.7: 128641241 - 128660998 on Build GRCh38
          Chromosome Location:
          Chr.11: 126987 - 131920 on Build GRCh38
          Chromosome Location:
          Chr.19: 197016 - 202209 on Build GRCh38
          Chromosome Location:
          Chr.4: 119405523 - 119410660 on Build GRCh38
          Chromosome Location:
          Chr.5: 181323506 - 181328195 on Build GRCh38
          Chromosome Location:
          -
          Chromosome Location:
          Chr.1: 490756 - 495445 on Build GRCh38
          Chromosome Location:
          Chr.1: 725759 - 730351 on Build GRCh38
          Chromosome Location:
          Chr.7: 63765043 - 63767827 on Build GRCh38
          Chromosome Location:
          Chr.7: 56809124 - 56812736 on Build GRCh38
          Chromosome Location:
          Chr.1: 134773 - 140566 on Build GRCh38
          UniGene ID:
          Hs.497951
          Hs.414183
          Hs.722350
          Hs.645398
          Hs.356758
          Hs.448518
          Hs.722350
          Hs.520384
          Hs.446409
          Hs.728864
          -
          Hs.457696
          Hs.534942
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          siRNA ID HSS167568
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price 459.00
          Your Price
          Online offer:
          459.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: LINC00999
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_024497.2 8 4682
          GenBank mRNA AK128746.1 - 2 3141
          BQ000710.1 - 1 585
          Gene Symbol: LINC00265
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_026999.1 12 4175
          GenBank mRNA AK097404.1 - 2 1719
          Gene Symbol: LINC01000
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_024368.1 5 3784
          GenBank mRNA AK294679.1 - 1 1512
          BC047304.1 - 1 754
          BC062355.1 - 6 2230
          BC131589.1 - 6 2222
          Gene Symbol: LINC01001
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_028326.1 3 4015
          Gene Symbol: LINC01002
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_028324.1 3 4386
          GenBank mRNA AK128780.1 - 2 1691
          BC117566.1 - 1 437
          Gene Symbol: LINC01061
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_037596.1 3 4223
          Gene Symbol: LOC100132062
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_028325.1 3 3771
          GenBank mRNA BC112924.1 - 2 2122
          Gene Symbol: LOC100132217
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AK128836.1 - 3 3427
          Gene Symbol: LOC100132287
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_028322.1 3 3771
          Gene Symbol: LOC100133331
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_028327.1 3 3674
          GenBank mRNA AK097080.1 - 4 1822
          Gene Symbol: LOC107986805
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_017012932.1 XP_016868421.1 1 1643
          Gene Symbol: LOC401357
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_130727.1 1 3147
          GenBank mRNA AK125790.1 - 1 3146
          AK128114.1 - 1 2888
          Gene Symbol: LOC729737
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_039983.2 3 4875

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0