Search
Search

FAM65C| Gene Symbol: FAM65C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290268.1 | NP_001277197.1 | 1 | 387 | ||
| NR_110890.1 | 1 | 387 | ||||
| XM_005260294.3 | XP_005260351.1 | 2 | 384 | |||
| XM_006723713.3 | XP_006723776.1 | 1 | 447 | |||
| XM_011528578.2 | XP_011526880.1 | 2 | 412 | |||
| XM_011528579.2 | XP_011526881.1 | 1 | 447 | |||
| XM_011528580.2 | XP_011526882.1 | 1 | 447 | |||
| XM_011528581.2 | XP_011526883.1 | 1 | 447 | |||
| XM_011528584.2 | XP_011526886.1 | 1 | 449 | |||
| XM_011528586.2 | XP_011526888.1 | 1 | 450 | |||
| XM_017027682.1 | XP_016883171.1 | 1 | 449 | |||
| GenBank mRNA | AK124347.1 | - | 1 | 399 | ||
| AK295781.1 | - | 1 | 78 | |||
| AK299337.1 | - | 1 | 236 | |||
| DC400584.1 | - | 1 | 388 | |||