Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HDAC6 SIRNA Assays ›
          Gene Symbol:
          HDAC6
          Gene Name:
          histone deacetylase 6
          Gene Aliases:
          CPBHM, HD6, JM21, PPP1R90
          Chromosome Location:
          Chr.X: 48801377 - 48824982 on Build GRCh38
          UniGene ID:
          Hs.6764
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS173319
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: HDAC6
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001321225.1 NP_001308154.1 27, 28 3454
          NM_001321226.1 NP_001308155.1 26, 27 3864
          NM_001321227.1 NP_001308156.1 26, 27 3585
          NM_001321228.1 NP_001308157.1 26, 27 3548
          NM_001321229.1 NP_001308158.1 26, 27 3402
          NM_006044.3 NP_006035.2 26, 27 3455
          NR_135591.1 24, 25 3135
          GenBank mRNA AB020708.1 - 26, 27 3370
          AF132609.1 - 25, 26 3298
          AF174499.1 - 20, 21 2852
          AJ011972.1 - 26, 27 3391
          AK001543.1 - 10, 11 1840
          AK095124.1 - 15, 16 2391
          AK122954.1 - 14, 15 2849
          AK127118.1 - 14, 15 4003
          AK131286.1 - 4, 5 1769
          AK302926.1 - 26, 27 3393
          AK303012.1 - 24, 25 3296
          AL137696.1 - 25, 26 4115
          BC013219.1 - 3, 4 950
          BC013737.1 - 25, 26 3280 2984860
          BC069243.1 - 26, 27 3397 6584427

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          protein polyubiquitination
          transcription, DNA-templated
          protein deacetylation
          misfolded or incompletely synthesized protein catabolic process
          intracellular protein transport
          organelle organization
          negative regulation of microtubule depolymerization
          response to toxic substance
          positive regulation of signal transduction
          response to organic substance
          regulation of receptor activity
          regulation of autophagy
          positive regulation of epithelial cell migration
          negative regulation of hydrogen peroxide metabolic process
          positive regulation of receptor biosynthetic process
          macroautophagy
          histone deacetylation
          regulation of protein stability
          lysosome localization
          peptidyl-lysine deacetylation
          cellular response to topologically incorrect protein
          regulation of gene expression, epigenetic
          ubiquitin-dependent protein catabolic process via the multivesicular body sorting pathway
          protein complex disassembly
          negative regulation of protein complex disassembly
          regulation of fat cell differentiation
          negative regulation of proteolysis
          negative regulation of transcription, DNA-templated
          negative regulation of oxidoreductase activity
          mitochondrion localization
          response to misfolded protein
          regulation of microtubule-based movement
          regulation of androgen receptor signaling pathway
          regulation of establishment of protein localization
          cellular response to hydrogen peroxide
          aggresome assembly
          polyubiquitinated misfolded protein transport
          Hsp90 deacetylation
          response to growth factor
          histone H3 deacetylation
          cellular response to misfolded protein
          positive regulation of chaperone-mediated protein complex assembly
          tubulin deacetylation
          mitophagy in response to mitochondrial depolarization
          positive regulation of hydrogen peroxide-mediated programmed cell death
          regulation of mitophagy
          core promoter binding
          actin binding
          histone deacetylase activity
          protein binding
          beta-catenin binding
          microtubule binding
          zinc ion binding
          enzyme binding
          polyubiquitin binding
          ubiquitin protein ligase binding
          NAD-dependent histone deacetylase activity (H3-K14 specific)
          histone deacetylase binding
          tubulin deacetylase activity
          alpha-tubulin binding
          tau protein binding
          beta-tubulin binding
          misfolded protein binding
          Hsp90 protein binding
          dynein complex binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0