Search
Search

TSPAN32| Gene Symbol: TSPAN32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139022.2 | NP_620591.3 | 1 | 181 | ||
| XM_005252719.4 | XP_005252776.1 | 1 | 175 | |||
| XM_005252720.4 | XP_005252777.1 | 1 | 175 | |||
| XM_011519815.2 | XP_011518117.2 | 1 | 172 | |||
| XM_011519818.2 | XP_011518120.2 | 1 | 172 | |||
| XM_011519819.1 | XP_011518121.1 | 1 | 56 | |||
| XM_011519822.1 | XP_011518124.1 | 1 | 175 | |||
| XM_011519828.2 | XP_011518130.1 | 1 | 47 | |||
| XM_011519829.2 | XP_011518131.2 | 1 | 172 | |||
| GenBank mRNA | AK128812.1 | - | 1 | 181 | ||
| AK307638.1 | - | 1 | 181 | |||
| AK310116.1 | - | 1 | 47 | |||
| AY039001.1 | - | 1 | 50 | |||
| BC016693.1 | - | 1 | 185 | 4071111 | ||
| BG506933.1 | - | 1 | 182 | |||