Search
Search

EDIL3| Gene Symbol: EDIL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005711.4 | NP_005702.3 | 2, 3 | 683 | ||
| GenBank mRNA | AK312858.1 | - | 2, 3 | 200 | ||
| BC030828.1 | - | 2, 3 | 609 | 4791845 | ||
| U70312.1 | - | 2, 3 | 300 | |||