Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CELF1 SIRNA Assays ›
Gene Symbol:
CELF1
Gene Name:
CUGBP, Elav-like family member 1
Gene Aliases:
BRUNOL2, CUG-BP, CUGBP, CUGBP1, EDEN-BP, NAB50, NAPOR, hNab50
Chromosome Location:
Chr.11: 47465937 - 47565569 on Build GRCh38
UniGene ID:
Hs.595333
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS173815
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CELF1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001025596.2
NP_001020767.1
10
1144
NM_001172639.1
NP_001166110.1
13
2106
NM_001172640.1
NP_001166111.1
10
1141
NM_006560.3
NP_006551.1
11
1381
NM_198700.2
NP_941989.1
11
1326
XM_011519848.2
XP_011518150.1
11
1219
XM_011519850.2
XP_011518152.2
11
1455
XM_011519852.2
XP_011518154.1
13
1567
XM_011519854.1
XP_011518156.1
15
1675
XM_011519855.1
XP_011518157.1
11
1396
XM_011519856.1
XP_011518158.1
12
1477
XM_011519857.1
XP_011518159.1
11
1348
XM_011519858.1
XP_011518160.1
11
1443
XM_011519859.1
XP_011518161.1
15
1663
XM_017017101.1
XP_016872590.1
14
1559
XM_017017102.1
XP_016872591.1
13
1478
XM_017017103.1
XP_016872592.1
12
1371
XM_017017104.1
XP_016872593.1
12
1371
XM_017017105.1
XP_016872594.1
12
1368
XM_017017106.1
XP_016872595.1
14
1556
XM_017017107.1
XP_016872596.1
12
1368
XM_017017108.1
XP_016872597.1
12
1359
XM_017017109.1
XP_016872598.1
11
1446
XM_017017110.1
XP_016872599.1
12
1359
XM_017017111.1
XP_016872600.1
12
1356
XM_017017112.1
XP_016872601.1
12
1356
XM_017017113.1
XP_016872602.1
12
1356
XM_017017114.1
XP_016872603.1
12
1356
XM_017017115.1
XP_016872604.1
13
1399
XM_017017116.1
XP_016872605.1
12
1434
XM_017017117.1
XP_016872606.1
11
1195
XM_017017118.1
XP_016872607.1
11
1317
XM_017017119.1
XP_016872608.1
13
1536
XM_017017120.1
XP_016872609.1
11
1348
XM_017017121.1
XP_016872610.1
11
1443
XM_017017122.1
XP_016872611.1
11
1348
XM_017017123.1
XP_016872612.1
11
1396
XM_017017124.1
XP_016872613.1
13
1487
XM_017017125.1
XP_016872614.1
11
1216
XM_017017126.1
XP_016872615.1
11
1345
XM_017017127.1
XP_016872616.1
11
1369
XM_017017128.1
XP_016872617.1
11
1393
XM_017017129.1
XP_016872618.1
12
1391
XM_017017130.1
XP_016872619.1
11
1345
XM_017017131.1
XP_016872620.1
11
1369
XM_017017132.1
XP_016872621.1
11
1393
XM_017017133.1
XP_016872622.1
11
1175
XM_017017134.1
XP_016872623.1
12
1314
XM_017017135.1
XP_016872624.1
11
1333
GenBank mRNA
AB210019.1
-
13
2106
AF248648.1
-
10
1152
AF267533.1
-
10
1145
AF267534.1
-
10
1136
AJ007988.1
-
10
1134
AK304430.1
-
10
1084
AK307679.1
-
5
606
BC031079.1
-
11
1326
5297141
U63289.1
-
11
1259
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
mRNA splice site selection
mRNA processing
germ cell development
spermatid development
embryo development
RNA interference
negative regulation of translation
positive regulation of multicellular organism growth
regulation of RNA splicing
nucleotide binding
translation repressor activity, nucleic acid binding
RNA binding
mRNA binding
protein binding
translation initiation factor binding
BRE binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0