Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ARPP21 SIRNA Assays ›
Gene Symbol:
ARPP21
Gene Name:
cAMP regulated phosphoprotein 21
Gene Aliases:
ARPP-21, R3HDM3, RCS, TARPP
Chromosome Location:
Chr.3: 35638981 - 35798933 on Build GRCh38
UniGene ID:
Hs.475902
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS173878
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ARPP21
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001025068.1
NP_001020239.1
5
527
NM_001025069.1
NP_001020240.1
5
536
NM_001267616.1
NP_001254545.1
5
1683
NM_001267617.1
NP_001254546.1
5
493
NM_001267618.1
NP_001254547.1
4
624
NM_001267619.1
NP_001254548.1
4
458
NM_016300.4
NP_057384.2
5
510
NM_198399.1
NP_938409.1
4
518
XM_005264811.2
XP_005264868.1
5
539
XM_006712943.2
XP_006713006.1
5
539
XM_006712944.2
XP_006713007.1
5
539
XM_011533298.2
XP_011531600.1
5
539
XM_011533299.2
XP_011531601.1
5
1083
XM_011533300.2
XP_011531602.1
5
702
XM_011533301.2
XP_011531603.1
5
533
XM_011533302.2
XP_011531604.1
6
625
XM_011533303.2
XP_011531605.1
6
1175
XM_017005574.1
XP_016861063.1
5
629
XM_017005575.1
XP_016861064.1
6
12262
XM_017005576.1
XP_016861065.1
5
1083
XM_017005577.1
XP_016861066.1
5
533
XM_017005578.1
XP_016861067.1
5
509
XM_017005579.1
XP_016861068.1
5
1083
XM_017005580.1
XP_016861069.1
6
1175
XM_017005581.1
XP_016861070.1
6
12262
XM_017005582.1
XP_016861071.1
5
1083
XM_017005583.1
XP_016861072.1
5
533
XM_017005584.1
XP_016861073.1
5
1083
XM_017005585.1
XP_016861074.1
5
1083
XM_017005586.1
XP_016861075.1
5
539
XM_017005587.1
XP_016861076.1
5
702
XM_017005588.1
XP_016861077.1
6
1175
XM_017005589.1
XP_016861078.1
6
625
XM_017005590.1
XP_016861079.1
6
12262
XM_017005591.1
XP_016861080.1
5
1083
XM_017005592.1
XP_016861081.1
5
1083
XM_017005593.1
XP_016861082.1
5
629
XM_017005594.1
XP_016861083.1
5
533
XM_017005595.1
XP_016861084.1
6
1175
XM_017005596.1
XP_016861085.1
5
1083
XM_017005597.1
XP_016861086.1
5
1083
XM_017005598.1
XP_016861087.1
6
1175
XM_017005599.1
XP_016861088.1
5
1083
XM_017005600.1
XP_016861089.1
5
702
XM_017005601.1
XP_016861090.1
5
533
XM_017005602.1
XP_016861091.1
5
1083
XM_017005603.1
XP_016861092.1
5
702
XM_017005604.1
XP_016861093.1
5
533
XM_017005605.1
XP_016861094.1
6
1175
XM_017005606.1
XP_016861095.1
6
625
XM_017005607.1
XP_016861096.1
5
1083
XM_017005608.1
XP_016861097.1
5
1083
XM_017005609.1
XP_016861098.1
6
1175
XM_017005610.1
XP_016861099.1
5
1083
XM_017005611.1
XP_016861100.1
5
1083
XM_017005612.1
XP_016861101.1
5
1083
GenBank mRNA
AF112220.1
-
4
483
AK055064.1
-
5
536
AK289649.1
-
4
473
AK289868.1
-
5
530
AK294484.1
-
4
471
AV718064.1
-
5
527
AV718166.1
-
5
536
BC017805.1
-
4
518
BC036399.1
-
5
498
5263894
BC041385.1
-
5
1271
BC051828.1
-
5
510
6012704
BC066651.1
-
3
1037
BX537647.1
-
1
640
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
biological_process
cellular response to heat
molecular_function
nucleic acid binding
calmodulin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0