Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PACSIN2 SIRNA Assays ›
          Gene Symbol:
          PACSIN2
          Gene Name:
          protein kinase C and casein kinase substrate in neurons 2
          Gene Aliases:
          SDPII
          Chromosome Location:
          Chr.22: 42869766 - 43015690 on Build GRCh38
          UniGene ID:
          Hs.162877
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS174200
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PACSIN2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001184970.1 NP_001171899.1 6, 7 1013
          NM_001184971.1 NP_001171900.1 6, 7 938
          NM_007229.3 NP_009160.2 6, 7 1000
          XM_005261319.3 XP_005261376.1 6, 7 1525
          XM_011529846.1 XP_011528148.1 6, 7 1525
          XM_011529847.2 XP_011528149.1 6, 7 1110
          XM_011529848.2 XP_011528150.1 6, 7 962
          XM_011529849.2 XP_011528151.1 6, 7 1146
          XM_011529850.2 XP_011528152.1 6, 7 1337
          XM_011529851.2 XP_011528153.1 6, 7 1027
          XM_011529852.1 XP_011528154.1 5, 6 1388
          XM_011529853.1 XP_011528155.1 6, 7 1525
          XM_017028562.1 XP_016884051.1 6, 7 991
          XM_017028563.1 XP_016884052.1 6, 7 1043
          XM_017028564.1 XP_016884053.1 6, 7 1101
          XM_017028565.1 XP_016884054.1 6, 7 1130
          XM_017028566.1 XP_016884055.1 6, 7 1166
          XM_017028567.1 XP_016884056.1 6, 7 1357
          XM_017028568.1 XP_016884057.1 5, 6 1388
          GenBank mRNA AL136845.1 - 6, 7 938
          BC008037.2 - 6, 7 1000 2967052
          CR456536.1 - 5, 6 850
          CR533523.1 - 5, 6 778

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          actin or actin-binding cytoskeletal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          actin cytoskeleton organization
          regulation of endocytosis
          protein localization to endosome
          negative regulation of endocytosis
          cell projection morphogenesis
          caveola assembly
          caveolin-mediated endocytosis
          membrane tubulation
          cell-cell adhesion
          transporter activity
          protein binding
          cytoskeletal protein binding
          lipid binding
          identical protein binding
          phosphatidic acid binding
          cadherin binding involved in cell-cell adhesion

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0