Search
Search

AHNAK2| Gene Symbol: AHNAK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138420.2 | NP_612429.2 | 7 | 15129 | ||
| XM_005267299.1 | XP_005267356.1 | 7 | 14982 | |||
| GenBank mRNA | AB095939.1 | - | 1 | 6029 | ||
| AK126472.1 | - | 1 | 1611 | |||
| AL834367.1 | - | 1 | 173 | |||
| BC090889.1 | - | 3 | 247 | |||
| BC144467.1 | - | 1 | 514 | |||
| BX537923.1 | - | 1 | 1011 | |||
| BX538040.1 | - | 1 | 785 | |||
| BX538106.1 | - | 1 | 1251 | |||
| BX538133.1 | - | 1 | 672 | |||