Search
Search

FAM161B| Gene Symbol: FAM161B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152445.2 | NP_689658.2 | 3 | 884 | ||
| XM_011536475.1 | XP_011534777.1 | 3 | 685 | |||
| GenBank mRNA | AK056259.1 | - | 3 | 602 | ||
| AK302982.1 | - | 3 | 884 | |||
| BC053909.1 | - | 3 | 556 | 6501484 | ||