Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CTNND1 SIRNA Assays ›
          Gene Symbol:
          CTNND1 TMX2-CTNND1
          Gene Name:
          catenin delta 1
          TMX2-CTNND1 readthrough (NMD candidate)
          Gene Aliases:
          CAS, CTNND, P120CAS, P120CTN, p120, p120(CAS), p120(CTN)
          -
          Chromosome Location:
          Chr.11: 57761762 - 57819180 on Build GRCh38
          Chromosome Location:
          Chr.11: 57712523 - 57819180 on Build GRCh38
          UniGene ID:
          Hs.166011
          Hs.166011
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID HSS175607
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CTNND1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001085458.1 NP_001078927.1 12, 13 2536
          NM_001085459.1 NP_001078928.1 11, 12 2518
          NM_001085460.1 NP_001078929.1 11, 12 2445
          NM_001085461.1 NP_001078930.1 11, 12 2518
          NM_001085462.1 NP_001078931.1 10, 11 2326
          NM_001085463.1 NP_001078932.1 10, 11 2055
          NM_001085464.1 NP_001078933.1 9, 10 2037
          NM_001085465.1 NP_001078934.1 9, 10 2037
          NM_001085466.1 NP_001078935.1 10, 11 2055
          NM_001085467.1 NP_001078936.1 9, 10 2110
          NM_001085468.1 NP_001078937.1 9, 10 2037
          NM_001085469.1 NP_001078938.1 8, 9 1965
          NM_001206883.1 NP_001193812.1 11, 12 2235
          NM_001206884.1 NP_001193813.1 11, 12 2235
          NM_001206885.1 NP_001193814.1 12, 13 2536
          NM_001206886.1 NP_001193815.1 10, 11 2217
          NM_001206887.1 NP_001193816.1 10, 11 2217
          NM_001206888.1 NP_001193817.1 10, 11 2217
          NM_001206889.1 NP_001193818.1 11, 12 2336
          NM_001206890.1 NP_001193819.1 9, 10 2059
          NM_001206891.1 NP_001193820.1 11, 12 2409
          NM_001331.2 NP_001322.1 11, 12 2518
          GenBank mRNA AF062317.1 - 11, 12 2483
          AF062318.1 - 7, 8 1259
          AF062319.1 - 11, 12 2200
          AF062320.1 - 11, 12 2200
          AF062321.1 - 12, 13 2501
          AF062322.1 - 10, 11 2182
          AF062323.1 - 10, 11 2020
          AF062324.1 - 11, 12 2410
          AF062325.1 - 10, 11 2182
          AF062326.1 - 6, 7 1241
          AF062327.1 - 10, 11 2182
          AF062328.1 - 11, 12 2483
          AF062329.1 - 11, 12 2301
          AF062330.1 - 9, 10 2024
          AF062331.1 - 11, 12 2374
          AF062332.1 - 9, 10 2002
          AF062333.1 - 6, 7 1241
          AF062334.1 - 9, 10 2075
          AF062335.1 - 9, 10 2002
          AF062336.1 - 8, 9 1930
          AF062337.1 - 10, 11 2020
          AF062338.1 - 9, 10 2002
          AF062339.1 - 11, 12 2483
          AF062340.1 - 6, 7 1314
          AF062341.1 - 12, 13 2501
          AF062342.1 - 7, 8 1259
          AF062343.1 - 10, 11 2291
          AF062344.1 - 6, 7 1241
          AK292554.1 - 11, 12 2460
          BC010501.1 - 7, 8 1639
          BC075795.1 - 11, 12 2334 5269008
          BT007333.1 - 5, 6 978
          CR749275.1 - 12, 13 2296
          Gene Symbol: TMX2-CTNND1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_037646.1 13, 14 2524
          GenBank mRNA AB002382.1 - 13, 14 2430

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          intermediate filament binding protein

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          cell adhesion
          brain development
          Wnt signaling pathway
          single organismal cell-cell adhesion
          adherens junction organization
          negative regulation of canonical Wnt signaling pathway
          receptor binding
          protein binding
          protein kinase binding
          cadherin binding
          cadherin binding involved in cell-cell adhesion

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0