Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other AP2B1 SIRNA Assays ›
          Gene Symbol:
          AP2B1
          Gene Name:
          adaptor related protein complex 2 beta 1 subunit
          Gene Aliases:
          ADTB2, AP105B, AP2-BETA, CLAPB1
          Chromosome Location:
          Chr.17: 35586899 - 35726417 on Build GRCh38
          UniGene ID:
          Hs.514819
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS176011
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: AP2B1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001030006.1 NP_001025177.1 8 1219
          NM_001282.2 NP_001273.1 8 1219
          XM_005257937.3 XP_005257994.1 8 1284
          XM_005257938.2 XP_005257995.1 8 1185
          XM_005257941.2 XP_005257998.1 8 1154
          XM_011524448.1 XP_011522750.1 8 1583
          XM_011524449.2 XP_011522751.1 8 1284
          XM_011524450.1 XP_011522752.1 8 1185
          XM_011524451.1 XP_011522753.1 8 1154
          XM_011524452.1 XP_011522754.1 9 1352
          XM_011524453.1 XP_011522755.1 9 1270
          XM_011524454.1 XP_011522756.1 9 1286
          XM_011524455.2 XP_011522757.1 9 1270
          XM_017024284.1 XP_016879773.1 8 1284
          XM_017024285.1 XP_016879774.1 9 1352
          XM_017024286.1 XP_016879775.1 9 1317
          XM_017024287.1 XP_016879776.1 8 1284
          GenBank mRNA AK289528.1 - 8 1160
          AK292531.1 - 8 1097
          AK301522.1 - 8 1054
          AK304126.1 - 4 574
          AY341427.1 - 9 1254
          BC006201.2 - 8 1126 3532472
          CR749392.1 - 8 1202
          M34175.1 - 8 1207

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          membrane traffic protein

          Gene Ontology Categories:

          Function(s) Process(es)

          ventricular septum development
          intracellular protein transport
          microtubule-based movement
          antigen processing and presentation of exogenous peptide antigen via MHC class II
          aorta development
          negative regulation of epidermal growth factor receptor signaling pathway
          ephrin receptor signaling pathway
          clathrin coat assembly
          regulation of defense response to virus by virus
          Wnt signaling pathway, planar cell polarity pathway
          coronary vasculature development
          clathrin-mediated endocytosis
          signal sequence binding
          protein binding
          protein transporter activity
          clathrin binding
          protein complex binding
          clathrin adaptor activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-rhr4g:80/100.66.78.163:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0