Search
Search

GALNTL5| Gene Symbol: GALNTL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145292.3 | NP_660335.2 | 4 | 686 | ||
| NR_033169.1 | 5 | 935 | ||||
| XM_006715862.3 | XP_006715925.1 | 5 | 761 | |||
| XM_011515849.2 | XP_011514151.1 | 3 | 373 | |||
| XM_011515852.2 | XP_011514154.1 | 3 | 509 | |||
| XM_011515853.2 | XP_011514155.1 | 2 | 267 | |||
| XM_017011793.1 | XP_016867282.1 | 4 | 1960 | |||
| XM_017011794.1 | XP_016867283.1 | 3 | 258 | |||
| XM_017011795.1 | XP_016867284.1 | 4 | 2061 | |||
| XM_017011796.1 | XP_016867285.1 | 4 | 2062 | |||
| GenBank mRNA | AK093091.1 | - | 5 | 888 | ||
| AK315667.1 | - | 5 | 895 | |||
| BC022021.2 | - | 4 | 639 | 4825619 | ||