Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DMBT1 SIRNA Assays ›
Gene Symbol:
DMBT1
Gene Name:
deleted in malignant brain tumors 1
Gene Aliases:
GP340, SAG, muclin
Chromosome Location:
Chr.10: 122560665 - 122643736 on Build GRCh38
UniGene ID:
Hs.279611
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS176205
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DMBT1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001320644.1
NP_001307573.1
49, 50
6273
NM_004406.2
NP_004397.2
36, 37
4392
NM_007329.2
NP_015568.2
49, 50
6276
NM_017579.2
NP_060049.2
48, 49
6246
XM_006717660.3
XP_006717723.1
51, 52
6603
XM_006717665.3
XP_006717728.1
50, 51
6555
XM_011539388.2
XP_011537690.1
52, 53
6651
XM_011539389.2
XP_011537691.1
52, 53
6648
XM_011539390.2
XP_011537692.1
51, 52
6621
XM_011539391.2
XP_011537693.1
51, 52
6621
XM_011539392.2
XP_011537694.1
51, 52
6621
XM_011539393.2
XP_011537695.1
51, 52
6621
XM_011539394.2
XP_011537696.1
51, 52
6621
XM_011539395.2
XP_011537697.1
51, 52
6621
XM_011539396.2
XP_011537698.1
51, 52
6603
XM_011539398.2
XP_011537700.1
50, 51
6588
XM_011539399.2
XP_011537701.1
50, 51
6555
XM_011539400.2
XP_011537702.1
51, 52
6327
XM_011539401.2
XP_011537703.1
49, 50
6264
XM_011539402.2
XP_011537704.1
49, 50
6264
XM_011539403.2
XP_011537705.1
49, 50
6264
XM_011539405.2
XP_011537707.1
49, 50
6265
XM_011539407.2
XP_011537709.1
49, 50
6258
XM_011539408.2
XP_011537710.1
49, 50
6258
XM_011539409.2
XP_011537711.1
49, 50
6258
XM_011539410.2
XP_011537712.1
49, 50
6258
XM_011539411.2
XP_011537713.1
48, 49
6234
XM_011539413.2
XP_011537715.1
46, 47
5870
XM_011539414.2
XP_011537716.1
36, 37
4371
XM_011539415.2
XP_011537717.1
34, 35
4325
XM_017015798.1
XP_016871287.1
37, 38
4913
GenBank mRNA
AF159456.1
-
49, 50
6276
AJ000342.1
-
36, 37
4392
AJ243212.1
-
49, 50
6273
AJ243224.1
-
48, 49
6246
AJ297935.1
-
49, 50
6257
AK304149.1
-
23, 24
2416
BC153299.1
-
33, 34
4010
40147087
BX640988.1
-
7, 8
1048
FJ234410.1
-
5, 6
552
Back To Top
More Information
Panther Classification:
Molecular Function -
transmembrane signal receptor
serine protease
Gene Ontology Categories:
Function(s)
Process(es)
pattern recognition receptor signaling pathway
receptor-mediated endocytosis
multicellular organism development
protein transport
epithelial cell differentiation
induction of bacterial agglutination
cellular protein metabolic process
innate immune response
defense response to virus
scavenger receptor activity
protein binding
signaling pattern recognition receptor activity
zymogen binding
calcium-dependent protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0