Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other AGTR2 SIRNA Assays ›
          Gene Symbol:
          AGTR2
          Gene Name:
          angiotensin II receptor type 2
          Gene Aliases:
          AT2, ATGR2, MRX88
          Chromosome Location:
          Chr.X: 116170705 - 116174972 on Build GRCh38
          UniGene ID:
          Hs.405348
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS176285
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: AGTR2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000686.4 NP_000677.2 3 679
          XM_011537533.1 XP_011535835.1 2 577
          GenBank mRNA AK313927.1 - 3 681
          BC095504.1 - 1 940 6971944
          CR541969.1 - 1 472
          CR738498.1 - 3 647
          U16957.1 - 3 612

          Back To Top

          More Information


          Set Membership:

          G-protein coupled receptor

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          blood vessel remodeling
          regulation of systemic arterial blood pressure by circulatory renin-angiotensin
          renin-angiotensin regulation of aldosterone production
          vasodilation by angiotensin involved in regulation of systemic arterial blood pressure
          brain renin-angiotensin system
          cellular sodium ion homeostasis
          inflammatory response
          cell surface receptor signaling pathway
          G-protein coupled receptor signaling pathway
          G-protein coupled receptor signaling pathway coupled to cGMP nucleotide second messenger
          nitric oxide mediated signal transduction
          brain development
          regulation of blood pressure
          positive regulation of cell proliferation
          negative regulation of heart rate
          negative regulation of norepinephrine secretion
          cerebellar cortex development
          negative regulation of cell growth
          negative regulation of icosanoid secretion
          positive regulation of phosphoprotein phosphatase activity
          intracellular signal transduction
          regulation of metanephros size
          exploration behavior
          positive regulation of renal sodium excretion
          aldosterone secretion
          angiotensin-activated signaling pathway
          dopamine biosynthetic process
          regulation of transcription factor import into nucleus
          negative regulation of blood vessel endothelial cell migration
          positive regulation of nitric oxide biosynthetic process
          positive regulation of transcription, DNA-templated
          positive regulation of vasodilation
          negative regulation of fibroblast proliferation
          positive regulation of cytokine secretion
          positive regulation of nitric-oxide synthase activity
          negative regulation of neurotrophin TRK receptor signaling pathway
          cell growth involved in cardiac muscle cell development
          cellular response to dexamethasone stimulus
          positive regulation of metanephric glomerulus development
          positive regulation of branching involved in ureteric bud morphogenesis
          extracellular negative regulation of signal transduction
          positive regulation of extrinsic apoptotic signaling pathway
          angiotensin type II receptor activity
          protein binding
          transcription factor binding
          peptide hormone binding
          receptor antagonist activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0