Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other EZH2 SIRNA Assays ›
Gene Symbol:
EZH2
Gene Name:
enhancer of zeste 2 polycomb repressive complex 2 subunit
Gene Aliases:
ENX-1, ENX1, EZH1, EZH2b, KMT6, KMT6A, WVS, WVS2
Chromosome Location:
Chr.7: 148807372 - 148884662 on Build GRCh38
UniGene ID:
Hs.444082
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS176652
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: EZH2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001203247.1
NP_001190176.1
9, 10
1165
NM_001203248.1
NP_001190177.1
9, 10
1138
NM_001203249.1
NP_001190178.1
9, 10
1265
NM_004456.4
NP_004447.2
9, 10
1180
NM_152998.2
NP_694543.1
8, 9
1048
XM_005249962.4
XP_005250019.1
9, 10
3606
XM_005249963.4
XP_005250020.1
9, 10
3579
XM_005249964.4
XP_005250021.1
9, 10
3580
XM_011515883.2
XP_011514185.1
10, 11
1155
XM_011515884.2
XP_011514186.1
9, 10
1287
XM_011515885.2
XP_011514187.1
9, 10
3596
XM_011515886.2
XP_011514188.1
10, 11
1304
XM_011515887.2
XP_011514189.1
9, 10
1153
XM_011515888.2
XP_011514190.1
9, 10
1275
XM_011515889.2
XP_011514191.1
9, 10
1077
XM_011515890.2
XP_011514192.1
8, 9
3506
XM_011515891.2
XP_011514193.1
7, 8
1240
XM_011515892.2
XP_011514194.1
9, 10
3624
XM_011515893.2
XP_011514195.1
8, 9
3489
XM_011515894.2
XP_011514196.1
8, 9
1005
XM_011515895.2
XP_011514197.1
8, 9
3479
XM_011515896.2
XP_011514198.1
8, 9
3490
XM_011515897.2
XP_011514199.1
10, 11
1331
XM_011515898.2
XP_011514200.1
10, 11
1258
XM_011515899.2
XP_011514201.1
9, 10
3626
XM_011515901.2
XP_011514203.1
9, 10
3626
XM_017011817.1
XP_016867306.1
9, 10
3622
XM_017011818.1
XP_016867307.1
10, 11
1289
XM_017011819.1
XP_016867308.1
9, 10
3607
XM_017011820.1
XP_016867309.1
8, 9
1036
XM_017011821.1
XP_016867310.1
10, 11
1316
GenBank mRNA
AK092676.1
-
8, 9
947
AK293239.1
-
9, 10
1076
AK302216.1
-
9, 10
1265
AK303585.1
-
9, 10
1103
AK314291.1
-
8, 9
990
BC010858.2
-
9, 10
1109
3901250
JF327843.1
-
7, 8
855
U61145.1
-
9, 10
1061
X95653.1
-
9, 10
1029
Back To Top
More Information
Panther Classification:
Molecular Function -
histone modifying enzyme
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
DNA methylation
chromatin organization
transcription, DNA-templated
regulation of transcription, DNA-templated
positive regulation of epithelial to mesenchymal transition
regulation of gliogenesis
skeletal muscle satellite cell maintenance involved in skeletal muscle regeneration
cerebellar cortex development
hippocampus development
response to estradiol
negative regulation of transcription elongation from RNA polymerase II promoter
regulation of cell proliferation
regulation of circadian rhythm
positive regulation of MAP kinase activity
negative regulation of sequence-specific DNA binding transcription factor activity
positive regulation of GTPase activity
negative regulation of epidermal cell differentiation
negative regulation of gene expression, epigenetic
negative regulation of transcription, DNA-templated
negative regulation of retinoic acid receptor signaling pathway
rhythmic process
negative regulation of striated muscle cell differentiation
cellular response to hydrogen peroxide
G1 to G0 transition
histone H3-K27 methylation
protein localization to chromatin
positive regulation of protein serine/threonine kinase activity
positive regulation of dendrite development
negative regulation of G1/S transition of mitotic cell cycle
core promoter binding
DNA binding
chromatin binding
RNA binding
protein binding
protein-lysine N-methyltransferase activity
histone-lysine N-methyltransferase activity
chromatin DNA binding
histone methyltransferase activity
ribonucleoprotein complex binding
sequence-specific DNA binding
histone methyltransferase activity (H3-K27 specific)
promoter-specific chromatin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0