Search
Search

AKR7A3| Gene Symbol: AKR7A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012067.2 | NP_036199.2 | 2 | 314 | ||
| XM_011541046.2 | XP_011539348.1 | 2 | 890 | |||
| XM_017000714.1 | XP_016856203.1 | 2 | 48 | |||
| GenBank mRNA | AF040639.1 | - | 2 | 321 | ||
| AJ271799.1 | - | 2 | 244 | |||
| BC025709.1 | - | 2 | 300 | 5224626 | ||
| BC031562.1 | - | 2 | 256 | 5184829 | ||
| BC042420.1 | - | 2 | 280 | 5186889 | ||
| BM920315.1 | - | 2 | 278 | |||