Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PDXDC1 SIRNA Assays ›
Gene Symbol:
PDXDC1
PDXDC2P
Gene Name:
pyridoxal dependent decarboxylase domain containing 1
pyridoxal dependent decarboxylase domain containing 2, pseudogene
Gene Aliases:
LP8165
NPIP, PDXDC2
Chromosome Location:
Chr.16: 14974591 - 15139410 on Build GRCh38
Chromosome Location:
Chr.16: 69976297 - 70065948 on Build GRCh38
UniGene ID:
Hs.370781
Hs.513695
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID
HSS177120
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
459.00
Your Price
Online offer:
459.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PDXDC1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001285444.1
NP_001272373.1
15
1511
NM_001285445.1
NP_001272374.1
15
1508
NM_001285447.1
NP_001272376.1
16
1404
NM_001285448.1
NP_001272377.1
15
1417
NM_001285449.1
NP_001272378.1
16
1592
NM_001324019.1
NP_001310948.1
16
1589
NM_001324020.1
NP_001310949.1
16
1589
NM_001324021.1
NP_001310950.1
15
1511
NM_015027.3
NP_055842.2
16
1592
XM_005255173.1
XP_005255230.1
16
1548
XM_005255176.2
XP_005255233.1
16
1474
XM_006720865.2
XP_006720928.2
15
1310
XM_017023059.1
XP_016878548.1
17
1484
XM_017023060.1
XP_016878549.1
16
1654
XM_017023061.1
XP_016878550.1
16
1424
XM_017023062.1
XP_016878551.1
16
1505
XM_017023063.1
XP_016878552.1
17
1484
XM_017023064.1
XP_016878553.1
15
1412
XM_017023065.1
XP_016878554.1
17
1484
GenBank mRNA
AK025504.1
-
3
226
AK090552.1
-
3
1634
AK295168.1
-
16
1397
AK299111.1
-
15
1342
AK299799.1
-
15
1402
AY203955.1
-
15
1337
BC025366.2
-
10
778
BC036520.1
-
16
1456
5270048
BC045554.1
-
16
1543
5295351
BC053946.1
-
16
1543
BC060871.1
-
16
1426
30347972
BX647809.1
-
15
1352
BX648066.1
-
15
1349
CD370048.1
-
2
526
D87438.1
-
16
1452
Gene Symbol: PDXDC2P
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NR_003610.1
16
1524
GenBank mRNA
AK292860.1
-
16
1394
AL833251.1
-
16
1524
BC063633.1
-
15
1396
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
carboxylic acid metabolic process
cell-cell adhesion
carboxy-lyase activity
pyridoxal phosphate binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-sg7j2:80/100.66.75.98:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0