Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FN1 SIRNA Assays ›
          Gene Symbol:
          FN1
          Gene Name:
          fibronectin 1
          Gene Aliases:
          CIG, ED-B, FINC, FN, FNZ, GFND, GFND2, LETS, MSF
          Chromosome Location:
          Chr.2: 215360440 - 215436167 on Build GRCh38
          UniGene ID:
          Hs.203717
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS177362
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: FN1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001306129.1 NP_001293058.1 1, 2 413
          NM_001306130.1 NP_001293059.1 1, 2 413
          NM_001306131.1 NP_001293060.1 1, 2 413
          NM_001306132.1 NP_001293061.1 1, 2 413
          NM_002026.3 NP_002017.1 1, 2 411
          NM_054034.2 NP_473375.2 1, 2 411
          NM_212474.2 NP_997639.1 1, 2 411
          NM_212476.2 NP_997641.1 1, 2 411
          NM_212478.2 NP_997643.1 1, 2 411
          NM_212482.2 NP_997647.1 1, 2 411
          XM_005246397.1 XP_005246454.1 1, 2 510
          XM_005246398.1 XP_005246455.1 1, 2 510
          XM_005246399.1 XP_005246456.1 1, 2 510
          XM_005246401.1 XP_005246458.1 1, 2 510
          XM_005246402.1 XP_005246459.1 1, 2 510
          XM_005246403.1 XP_005246460.1 1, 2 510
          XM_005246404.1 XP_005246461.1 1, 2 510
          XM_005246405.1 XP_005246462.1 1, 2 510
          XM_005246406.1 XP_005246463.1 1, 2 510
          XM_005246407.1 XP_005246464.1 1, 2 510
          XM_005246408.1 XP_005246465.1 1, 2 510
          XM_005246409.1 XP_005246466.1 1, 2 510
          XM_005246410.1 XP_005246467.1 1, 2 510
          XM_005246411.1 XP_005246468.1 1, 2 510
          XM_005246412.1 XP_005246469.1 1, 2 510
          XM_005246414.1 XP_005246471.1 1, 2 510
          XM_005246416.1 XP_005246473.1 1, 2 510
          XM_017003692.1 XP_016859181.1 1, 2 510
          XM_017003693.1 XP_016859182.1 1, 2 510
          XM_017003694.1 XP_016859183.1 1, 2 510
          XM_017003695.1 XP_016859184.1 1, 2 510
          GenBank mRNA AB191261.1 - 1, 2 411
          AB209840.1 - 1, 2 403
          AJ276395.1 - 1, 2 201
          AJ535086.1 - 1, 2 201
          AJ849445.1 - 1, 2 201
          AL832202.1 - 1, 2 412
          AL832771.1 - 1, 2 412
          BC117176.1 - 1, 2 391 40125727
          BC143754.1 - 1, 2 391 9052268
          BC143763.1 - 1, 2 391 9052277
          BX537590.1 - 1, 2 412
          BX538017.1 - 1, 2 412
          BX538018.1 - 1, 2 412
          BX538045.1 - 1, 2 451
          BX640608.1 - 1, 2 411
          BX640731.1 - 1, 2 412
          BX640802.1 - 1, 2 414
          BX640803.1 - 1, 2 411
          BX640875.1 - 1, 2 412
          BX649182.1 - 1, 2 414
          CR749281.1 - 1, 2 450
          CR749316.1 - 1, 2 410
          CR749317.1 - 1, 2 410
          DA851446.1 - 1, 2 413
          X02761.1 - 1, 2 65

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          extracellular matrix structural protein

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          cell activation
          regulation of protein phosphorylation
          platelet degranulation
          response to ischemia
          acute-phase response
          cell-substrate junction assembly
          cell adhesion
          calcium-independent cell-matrix adhesion
          positive regulation of cell proliferation
          glial cell migration
          regulation of cell shape
          response to wounding
          response to ozone
          positive regulation of gene expression
          positive regulation of peptidase activity
          peptide cross-linking
          extracellular matrix disassembly
          extracellular matrix organization
          integrin activation
          substrate adhesion-dependent cell spreading
          cellular response to vascular endothelial growth factor stimulus
          endodermal cell differentiation
          cellular response to platelet-derived growth factor stimulus
          wound healing
          negative regulation of apoptotic process
          positive regulation of axon extension
          positive regulation of fibroblast proliferation
          leukocyte migration
          positive regulation of chemotaxis
          response to glucocorticoid
          regulation of ERK1 and ERK2 cascade
          cellular response to lipopolysaccharide
          cellular response to mercury ion
          cellular response to glucose stimulus
          cellular response to interleukin-1
          cellular response to prostaglandin E stimulus
          cellular response to transforming growth factor beta stimulus
          cellular response to BMP stimulus
          positive regulation of substrate-dependent cell migration, cell attachment to substrate
          negative regulation of transforming growth factor-beta secretion
          protease binding
          integrin binding
          protein binding
          collagen binding
          heparin binding
          peptidase activator activity
          identical protein binding
          mercury ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gqkn5:80/100.66.76.113:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0