Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MAP7D2 SIRNA Assays ›
Gene Symbol:
MAP7D2
Gene Name:
MAP7 domain containing 2
Gene Aliases:
-
Chromosome Location:
Chr.X: 20006713 - 20116996 on Build GRCh38
UniGene ID:
Hs.127951
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS177789
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MAP7D2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001168465.1
NP_001161937.1
7
919
NM_001168466.1
NP_001161938.1
5
661
NM_001168467.1
NP_001161939.1
6
633
NM_152780.3
NP_689993.2
6
796
XM_005274478.2
XP_005274535.1
7
782
XM_005274479.2
XP_005274536.1
7
749
XM_005274480.3
XP_005274537.1
6
683
XM_006724481.2
XP_006724544.1
8
929
XM_006724482.2
XP_006724545.1
7
806
XM_011545483.1
XP_011543785.1
8
896
XM_011545484.1
XP_011543786.1
8
879
XM_011545485.1
XP_011543787.1
8
814
XM_011545486.2
XP_011543788.1
8
929
XM_017029393.1
XP_016884882.1
7
773
XM_017029394.1
XP_016884883.1
7
756
XM_017029395.1
XP_016884884.1
7
732
XM_017029396.1
XP_016884885.1
7
691
XM_017029397.1
XP_016884886.1
7
667
XM_017029398.1
XP_016884887.1
6
683
GenBank mRNA
AK027409.1
-
6
698
AK127056.1
-
4
335
AK294788.1
-
6
627
AK296319.1
-
5
572
AK316117.1
-
5
569
BC037165.2
-
4
463
BC089400.1
-
7
840
3063898
BC136379.1
-
6
713
9020366
KP849473.1
-
7
754
KP849474.1
-
7
721
KP979782.1
-
7
788
KP979783.1
-
7
755
KP979784.1
-
7
812
KP979785.1
-
7
779
KP979786.1
-
7
812
Back To Top
More Information
Panther Classification:
Molecular Function -
non-motor microtubule binding protein
Gene Ontology Categories:
Function(s)
Process(es)
microtubule cytoskeleton organization
structural molecule activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-qfdf2:80/100.66.76.55:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline