Search
Search

SUN3| Gene Symbol: SUN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001030019.1 | NP_001025190.1 | 9, 10 | 1143 | ||
| NM_001284350.1 | NP_001271279.1 | 10, 11 | 1241 | |||
| NM_152782.3 | NP_689995.3 | 10, 11 | 1059 | |||
| XM_017011930.1 | XP_016867419.1 | 10, 11 | 1049 | |||
| GenBank mRNA | AF429967.1 | - | 9, 10 | 1134 | ||
| AI222206.1 | - | 1, 2 | 307 | |||
| AK302011.1 | - | 10, 11 | 1170 | |||
| BC026189.2 | - | 10, 11 | 1047 | 4819018 | ||
| BU623138.1 | - | 1, 2 | 306 | |||
| DQ099386.1 | - | 7, 8 | 698 | |||