Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PRPF40B SIRNA Assays ›
          Gene Symbol:
          PRPF40B FMNL3
          Gene Name:
          pre-mRNA processing factor 40 homolog B
          formin like 3
          Gene Aliases:
          HYPC
          FHOD3, FRL2, WBP-3, WBP3
          Chromosome Location:
          Chr.12: 49622717 - 49644669 on Build GRCh38
          Chromosome Location:
          Chr.12: 49636499 - 49707414 on Build GRCh38
          UniGene ID:
          Hs.706827
          Hs.179838
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID HSS177816
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PRPF40B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001031698.2 NP_001026868.2 18 1929
          NM_012272.2 NP_036404.1 17 2219
          XM_006719324.3 XP_006719387.1 17 2983
          XM_006719325.3 XP_006719388.1 17 2983
          XM_011538135.2 XP_011536437.1 17 2983
          XM_011538136.2 XP_011536438.1 17 2983
          XM_011538137.2 XP_011536439.1 16 2902
          XM_011538138.2 XP_011536440.1 16 2872
          XM_011538139.2 XP_011536441.1 18 1923
          XM_011538140.2 XP_011536442.1 18 1923
          XM_011538141.1 XP_011536443.1 17 1749
          XM_011538143.2 XP_011536445.1 17 1765
          XM_011538144.2 XP_011536446.1 16 1638
          XM_017019135.1 XP_016874624.1 17 1686
          XM_017019136.1 XP_016874625.1 16 1638
          XM_017019137.1 XP_016874626.1 16 1638
          XM_017019138.1 XP_016874627.1 17 2983
          XM_017019139.1 XP_016874628.1 13 1649
          GenBank mRNA AK027117.1 - 3 210
          AL137459.1 - 4 432
          AL834216.1 - 17 2219
          BC031917.1 - 15 1525
          BC050398.2 - 18 1765
          BC067364.1 - 17 1750 6644672
          Gene Symbol: FMNL3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_005269218.2 XP_005269275.1 27 11414
          XM_011538968.2 XP_011537270.1 27 11904
          XM_011538969.2 XP_011537271.1 26 11788
          XM_011538970.2 XP_011537272.1 27 11903
          XM_011538971.2 XP_011537273.1 27 11479
          XM_011538972.2 XP_011537274.1 26 11749
          XM_011538973.2 XP_011537275.2 23 10894
          XM_011538974.2 XP_011537276.1 28 11236

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          mRNA splicing, via spliceosome
          angiogenesis
          cytoskeleton organization
          regulation of cell shape
          cell migration
          actin cytoskeleton organization
          RNA binding
          actin binding
          Rho GTPase binding
          GTPase activating protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-69rxj:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0