Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GAK SIRNA Assays ›
Gene Symbol:
GAK
Gene Name:
cyclin G associated kinase
Gene Aliases:
DNAJ26, DNAJC26
Chromosome Location:
Chr.4: 849275 - 932390 on Build GRCh38
UniGene ID:
Hs.369607
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS177841
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: GAK
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001318134.1
NP_001305063.1
15, 16
2010
NM_005255.3
NP_005246.2
18, 19
2247
XM_005272268.2
XP_005272325.1
18, 19
2187
XM_005272270.2
XP_005272327.1
18, 19
2187
XM_011513425.2
XP_011511727.1
18, 19
2187
XM_011513426.2
XP_011511728.1
18, 19
2187
XM_011513427.2
XP_011511729.1
17, 18
2097
XM_011513428.2
XP_011511730.1
18, 19
2187
XM_011513429.2
XP_011511731.1
16, 17
2509
XM_011513430.1
XP_011511732.1
15, 16
1934
XM_011513431.2
XP_011511733.1
18, 19
2187
XM_011513432.2
XP_011511734.1
18, 19
2158
XM_011513434.2
XP_011511736.1
16, 17
1999
XM_017007991.1
XP_016863480.1
17, 18
2097
XM_017007992.1
XP_016863481.1
14, 15
1844
XM_017007993.1
XP_016863482.1
16, 17
1936
XM_017007994.1
XP_016863483.1
18, 19
2187
XM_017007995.1
XP_016863484.1
17, 18
2006
GenBank mRNA
AB208854.1
-
15, 16
1733
AK299554.1
-
15, 16
1882
AK309413.1
-
15, 16
1866
BC085005.1
-
15, 16
1993
6598984
D88435.1
-
18, 19
2044
Back To Top
More Information
Panther Classification:
Molecular Function -
membrane traffic protein
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
receptor-mediated endocytosis
endoplasmic reticulum organization
Golgi organization
cell cycle
negative regulation of neuron projection development
clathrin coat disassembly
clathrin-mediated endocytosis
establishment of protein localization to Golgi
establishment of protein localization to plasma membrane
Golgi to lysosome transport
protein serine/threonine kinase activity
protein binding
ATP binding
cyclin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0