Search
Search

PCDHB5| Gene Symbol: PCDHB5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015669.4 | NP_056484.2 | 1 | 475 | ||
| GenBank mRNA | AB209393.1 | - | 1 | 430 | ||
| AF134474.1 | - | 1 | 314 | |||
| AF152498.1 | - | 1 | 246 | |||
| AF217753.1 | - | 1 | 246 | |||
| AK023190.1 | - | 1 | 463 | |||
| BC001186.1 | - | 1 | 453 | 3528826 | ||
| BP310685.1 | - | 1 | 475 | |||
| DA389074.1 | - | 1 | 475 | |||